    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • LMCD1 Antibodies
          Proteintech
          LMCD1 Monoclonal Antibody (2A7B3), CoraLite® Plus 488
          View all (13) LMCD1 antibodies
          Tech Support
          Cite LMCD1 Monoclonal Antibody (2A7B3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          LMCD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          LMCD1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          LMCD1 Antibody (CL488-67834) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed MCF-7 cells using CoraLite® Plus 488 LMCD1 antibody (CL488-67834, Clone: 2A7B3) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          LMCD1 Antibody in Immunocytochemistry (ICC/IF)
          LMCD1 Antibody in Flow Cytometry (Flow)
          Product Details
          CL488-67834
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2A7B3
          Immunogen
          LMCD1 fusion protein Ag28331
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAKVAKDLN PGVKKMSLGQ LQSARGVACL GCKGTCSGFE PHSWRKICKS CKCSQEDHCL TSDLEDDRKI GRLLMDSKYS TLTARVKGGD GIRIYKRNRM IMTNPIATGK DPTFDTITYE WAPPGVTQKL GLQYMELIPK EKQPVTGTEG AFYRRRQLMH QLPIYDQDPS RCRGLLENEL KLMEEFVKQY KSEALGVGEV ALPGQGGLPK EEGKQQEKPE GAETTAATTN GSLSDPSKEV EYVCELCKGA APPDSPVVYS DRAGYNKQWH PTCFVCAKCS EPLVDLIYFW KDGAPWCGRH YCESLRPRCS GCDEIIFAED YQRVEDLAWH RKHFVCEGCE QLLSGRAYIV TKGQLLCPTC SKSKRS
          Target Information
          LMCD1 encodes a member of the LIM-domain family of zinc finger proteins. The encoded protein contains an N-terminal cysteine-rich domain and two C-terminal LIM domains. The presence of LIM domains suggests involvement in protein-protein interactions. The protein may act as a co-regulator of transcription along with other transcription factors. Alternate splicing results in multiple transcript variants of this gene.
          Bioinformatics
          Protein Aliases: Dyxin; LIM and cysteine-rich domains protein 1; LIM domain protein; unnamed protein product
          View more View less
          Gene Aliases: LMCD1
          View more View less
          UniProt ID: (Human) Q9NZU5
          View more View less
          Entrez Gene ID: (Human) 29995
          View more View less
          Function(s)
          transcription corepressor activity protein binding zinc ion binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of cardiac muscle hypertrophy positive regulation of calcineurin-NFAT signaling cascade
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline