    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • SHMT1 Antibodies
          Proteintech
          SHMT1 Monoclonal Antibody (2D5E6), CoraLite® Plus 488
          View all (13) SHMT1 antibodies
          Tech Support
          Cite SHMT1 Monoclonal Antibody (2D5E6), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          SHMT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SHMT1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          SHMT1 Antibody (CL488-67963) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 SHMT1 antibody (CL488-67963, Clone: 2D5E6) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SHMT1 Antibody in Immunocytochemistry (ICC/IF)
          SHMT1 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-67963
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Pig, Rabbit
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2D5E6
          Immunogen
          SHMT1 fusion protein Ag5833
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: PHGRIMGLD LPDGGHLTHG FMTDKKKISA TSIFFESMPY KVNPDTGYIN YDQLEENARL FHPKLIIAGT SCYSRNLEYA RLRKIADENG AYLMADMAHI SGLVAAGVVP SPFEHCHVVT TTTHKTLRGC RAGMIFYRKG VKSVDPKTGK EILYNLESLI NSAVFPGLQG GPHNHAIAGV AVALKQAMTL EFKVYQHQVV ANCRALSEAL TELGYKIVTG GSDNHLILVD LRSKGTDGGR AEKVLEACSI ACNKNTCPGD RSALRPSGLR LGTPALTSRG LLEKDFQKVA HFIHRGIELT LQIQSDTGVR ATLKEFKERL AGDKYQAAVQ ALREEVESFA SLFPLPGLPD F
          Target Information
          This gene encodes the cellular form of serine hydroxymethyltransferase, a pyridoxal phosphate-containing enzyme that catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon units for synthesis of methionine, thymidylate, and purines in the cytoplasm. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing of this gene results in 2 transcript variants encoding 2 different isoforms. Additional transcript variants have been described, but their biological validity has not been determined.
          Bioinformatics
          Protein Aliases: cytoplasmic serine hydroxymethyltransferase; cytosolic serine hydroxymethyltransferase; Glycine hydroxymethyltransferase; L-allo-threonine/L-threonine aldolase SHMT1; MGC15229; MGC24556; serine hydroxymethyltransferase 1 (soluble); serine hydroxymethyltransferase 2 (mitochondrial); Serine hydroxymethyltransferase, cytosolic; Serine methylase; SHMT; unnamed protein product
          View more View less
          Gene Aliases: CSHMT; MEL-32; SHMT; SHMT1
          View more View less
          UniProt ID: (Human) P34896, (Rabbit) P07511
          View more View less
          Entrez Gene ID: (Human) 6470, (Pig) 397181, (Rabbit) 100009405
          View more View less
          Function(s)
          translation repressor activity, nucleic acid binding glycine hydroxymethyltransferase activity protein binding transferase activity aldehyde-lyase activity pyridoxal phosphate binding small molecule binding identical protein binding protein homodimerization activity mRNA 5'-UTR binding serine binding methyltransferase
          Process(es)
          dTMP biosynthetic process regulation of translation glycine metabolic process L-serine metabolic process L-serine catabolic process one-carbon metabolic process purine nucleobase biosynthetic process negative regulation of translation glycine biosynthetic process from serine tetrahydrofolate interconversion carnitine biosynthetic process tetrahydrofolate metabolic process folic acid metabolic process protein homotetramerization cellular response to tetrahydrofolate cellular response to leukemia inhibitory factor
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline