    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Asparagine Synthetase Antibodies
          Proteintech
          ASNS Monoclonal Antibody (1C3D3), CoraLite® Plus 488
          View all (27) Asparagine Synthetase antibodies
          Tech Support
          Cite ASNS Monoclonal Antibody (1C3D3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          ASNS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ASNS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          ASNS Antibody (CL488-68004) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed SH-SY5Y cells using CoraLite Plus 488 ASNS antibody (CL488-68004, Clone: 1C3D3 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ASNS Antibody in Immunocytochemistry (ICC/IF)
          ASNS Antibody in Western Blot (WB)
          ASNS Monoclonal Antibody (1C3D3), CoraLite® Plus 488
          Product Details
          CL488-68004
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:4,000
          -
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1C3D3
          Immunogen
          ASNS fusion protein Ag6817
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: LMTDRRIGCL LSGGLDSSLV AATLLKQLKE AQVQYPLQTF AIGMEDSPDL LAARKVADHI GSEHYEVLFN SEEGIQALDE VIFSLETYDI TTVRASVGMY LISKYIRKNT DSVVIFSGEG SDELTQGYIY FHKAPSPEKA EEESERLLRE LYLFDVLRAD RTTAAHGLEL RVPFLDHRFS SYYLSLPPEM RIPKNGIEKH LLRETFEDSN LIPKEILWRP KEAFSDGITS VKNSWFKILQ EYVEHQVDDA MMANAAQKFP FNTPKTKEGY YYRQVFERHY PGRADWLSHY WMPKWINATD PSARTLTHYK SAVKA
          Target Information
          ASNS (asparagine synthetase [glutamine-hydrolyzing]) is involved in the subpathway that synthesizes f L-asparagine from L-aspartate. Mutations affecting the gene can result in asparagine synthetase deficiency (ASNSD).
          Bioinformatics
          Protein Aliases: asparagine synthetase; Asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11 cell cycle control protein; unnamed protein product
          View more View less
          Gene Aliases: ASNS; ASNSD; TS11
          View more View less
          UniProt ID: (Human) P08243
          View more View less
          Entrez Gene ID: (Human) 440
          View more View less
          Function(s)
          nucleotide binding asparagine synthase (glutamine-hydrolyzing) activity protein binding ATP binding ligase activity
          Process(es)
          asparagine biosynthetic process cellular amino acid biosynthetic process cellular response to glucose starvation negative regulation of apoptotic process positive regulation of mitotic cell cycle L-asparagine biosynthetic process
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline