    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • SMS Antibodies
          Proteintech
          SMS Monoclonal Antibody (1G8E9), CoraLite® Plus 488
          View all (20) SMS antibodies
          Tech Support
          Cite SMS Monoclonal Antibody (1G8E9), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SMS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SMS Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SMS Antibody (CL488-68040) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed HeLa cells using CoraLite® Plus 488 SMS antibody (CL488-68040, Clone: 1G8E9) at dilution of 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SMS Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-68040
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1G8E9
          Immunogen
          SMS fusion protein Ag8840
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAAARHSTL DFMLGAKADG ETILKGLQSI FQEQGMAESV HTWQDHGYLA TYTNKNGSFA NLRIYPHGLV LLDLQSYDGD AQGKEEIDSI LNKVEERMKE LSQDSTGRVK RLPPIVRGGA IDRYWPTADG RLVEYDIDEV VYDEDSPYQN IKILHSKQFG NILILSGDVN LAESDLAYTR AIMGSGKEDY TGKDVLILGG GDGGILCEIV KLKPKMVTMV EIDQMVIDGC KKYMRKTCGD VLDNLKGDCY QVLIEDCIPV LKRYAKEGRE FDYVINDLTA VPISTSPEED STWEFLRLIL DLSMKVLKQD GKYFTQGNCV NLTEALSLYE EQLGRLYCPV EFSKEIVCVP SYLELWVFYT VWKKAKP
          Target Information
          Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
          Bioinformatics
          Protein Aliases: Spermidine aminopropyltransferase; Spermine synthase; SPMSY; unnamed protein product
          View more View less
          Gene Aliases: MRSR; MRXSSR; SMS; SPMSY; SpS; SRS
          View more View less
          UniProt ID: (Human) P52788
          View more View less
          Entrez Gene ID: (Human) 6611
          View more View less
          Function(s)
          transferase activity spermine synthase activity
          Process(es)
          methionine metabolic process polyamine metabolic process polyamine biosynthetic process spermine biosynthetic process spermine metabolic process
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline