Immunogen sequence: DAQHESGQS SSRGSSHCDS PSPQEDGQIM FDVEMHTSRD HSSQSEEEVV EGEKEVEALK KSADWVSDWS S
This gene encodes a protein that interacts with the small GTPase rab11. A similar protein in rat binds the GTP-containing active form of rab11. This protein may play a role in endosome recycling. Alternate splicing results in multiple transcript variants.
Protein Aliases: Adenovirus E1B19K-binding protein B5; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; BCL2/adenovirus E1B 19-kd protein-interacting protein 3a; BCL2/adenovirus E1B 19kDa interacting protein 3 like; BNIP3a; similar to BNIP3; BNIP3L; NIP-3-like protein X; NIP3-like protein X; NIP3L; similar to Nip3; unnamed protein product
Gene Aliases: BNIP3A; BNIP3H; BNIP3L; NIP3L; NIX
UniProt ID: (Human) O60238
Entrez Gene ID: (Human) 665