    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Dystrophin Antibodies
          Proteintech
          Dystrophin Monoclonal Antibody (1G12E6), CoraLite® Plus 488
          View all (36) Dystrophin antibodies
          Tech Support
          Cite Dystrophin Monoclonal Antibody (1G12E6), CoraLite® Plus 488
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Dystrophin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Dystrophin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Dystrophin Antibody (CL488-68120) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed H9C2 cells using CoraLite® Plus 488 Dystrophin antibody (CL488-68120, Clone: 1G12E6) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Dystrophin Antibody in Immunocytochemistry (ICC/IF)
          Dystrophin Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Dystrophin Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL488-68120
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1:200
          -
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1G12E6
          Immunogen
          Dystrophin fusion protein Ag4392
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: YPMVEYCTP TTSGEDVRDF AKVLKNKFRT KRYFAKHPRM GYLPVQTVLE GDNMETPVTL INFWPVDSAP ASSPQLSHDD THSRIEHYAS RLAEMENSNG SYLNDSISPN ESIDDEHLLI QHYCQSLNQD SPLSQPRSPA QILISLESEE RGELERILAD LEEENRNLQA EYDRLKQQHE HKGLSPLPSP PEMMPTSPQS PRDAELIAEA KLLRQHKGRL EARMQILEDH NKQLESQLHR LRQLLEQPQA EAKVNGTTVS SPSTSLQRSD SSQPMLLRVV GSQTSDSMGE EDLLSPPQDT STGLEEVMEQ LNNSFPSSRG HNVGSLFHMA DDLGRAMESL VSVMTDEEGA E
          Target Information
          Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immunohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.
          Bioinformatics
          Protein Aliases: apodystrophin-3; apodystrophin-I; DMD; Dpm234; Duchenne muscular dystrophy (DMD); dystrophin; dystrophin delta-E40e; dystrophin Dp427-E40e; dystrophin Dp71d(delta71,73-74); dystrophin Dp71f-delta71,74; dystrophin E40e; dystrophin E51b; dystrophin, muscular dystrophy; dystrophin-related; GS1-19O24.1; Muscular dystrophy Duchenne and Becker types; mutant dystrophin; OTTHUMP00000215592; unnamed protein product; X-linked muscular dystrophy
          View more View less
          Gene Aliases: BMD; CMD3B; DNADMD1; Dp427; Dp71; DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; DXSmh7; DXSmh9; dys; mdx; MRX85; pke
          View more View less
          UniProt ID: (Human) P11532, (Rat) P11530, (Mouse) P11531
          View more View less
          Entrez Gene ID: (Human) 1756, (Rat) 24907, (Mouse) 13405
          View more View less
          Function(s)
          dystroglycan binding actin binding integrin binding structural constituent of cytoskeleton protein binding lamin binding zinc ion binding structural constituent of muscle myosin binding vinculin binding PDZ domain binding macromolecular complex binding nitric-oxide synthase binding
          Process(es)
          positive regulation of cell-matrix adhesion regulation of heart rate regulation of transcription, DNA-templated apoptotic process striated muscle contraction inflammatory response muscle organ development skeletal muscle tissue development establishment of blood-nerve barrier protein localization positive regulation of cell proliferation determination of adult lifespan response to xenobiotic stimulus gene expression regulation of gene expression regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion positive regulation of neuron projection development regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion regulation of skeletal muscle contraction response to denervation involved in regulation of muscle adaptation myotube cell development olfactory nerve structural organization cerebral cortex development cell differentiation neuron differentiation multicellular organism growth response to muscle stretch regulation of membrane potential muscle cell differentiation peptide biosynthetic process skeletal muscle tissue regeneration negative regulation of neuron differentiation positive regulation of neuron differentiation muscle cell cellular homeostasis neuron development neuron projection morphogenesis nucleus localization muscle cell development striated muscle cell development cardiac muscle contraction limb development bone development establishment of glial blood-brain barrier connective tissue development protein-containing complex assembly negative regulation of ERK1 and ERK2 cascade cardiac muscle cell action potential regulation of muscle system process regulation of cellular response to growth factor stimulus walking behavior synaptic signaling regulation of sodium ion transmembrane transport regulation of calcium ion transmembrane transport reactive oxygen species biosynthetic process muscle contraction cytoskeleton organization maintenance of blood-brain barrier camera-type eye development regulation of system process motile cilium assembly
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-l699q:80/100.66.79.160:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline