Immunogen sequence: YDLQESNVR LKLTIVSTVG FGDQINKEDS YKPIVEFIDA QFEAYLQEEL KIRRVLHTYH DSRIHVCLYF IAPTGHSLKS LDLVTMKKLD SKVNIIPIIA KADAISKSEL TKFKIKITSE LVSNGVQIYQ FPTDDESVAE INGTMNAHLP FAVIGSTEEL KIGNKMMRAR QYPWGTVQVE NEAHCDFVKL REMLIRVNME DLREQTHTRH YELYRRCKLE EMGFKDTDPD SKPFSLQETY EAKRNEFLGE LQKKEEEMRQ MFVQRVKEKE AELKEAEKEL HEKFDRLKKL HQDEKKKLED KKKSLDDEVN AFKQRKTAAE LLQSQGSQAG GSQTLKRDKE KKNNPWLCTE
SEPT6 is a member of the septin family of GTPases. Members of this family are required for cytokinesis. This gene is a member of the septin family of GTPases. Members of this family are required for cytokinesis. One version of pediatric acute myeloid leukemia is the result of a reciprocal translocation between chromosomes 11 and X, with the breakpoint associated with the genes encoding the mixed-lineage leukemia and septin 2 proteins. This gene encodes four transcript variants encoding three distinct isoforms. An additional transcript variant has been identified, but its biological validity has not been determined.
Protein Aliases: 2-Sep; 6-Sep; KIAA0128; MGC16619; MGC20339; RP5-876A24.2; septin 2; Septin-6; transcript variant VI; unnamed protein product
Gene Aliases: 2810035H17Rik; C920001C06Rik; KIAA0128; mKIAA0128; SEP2; Sep6; SEPT2; SEPT6; Septin-6; SEPTIN6
UniProt ID: (Human) Q14141, (Mouse) Q9R1T4
Entrez Gene ID: (Human) 23157, (Rabbit) 100341047, (Mouse) 56526, (Rat) 691335