    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PPA1 Antibodies
          Proteintech
          PPA1 Monoclonal Antibody (1D11F4), CoraLite® Plus 488
          View all (17) PPA1 antibodies
          Questions & Answers
          Cite PPA1 Monoclonal Antibody (1D11F4), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          PPA1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PPA1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          PPA1 Antibody (CL488-68178) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed HeLa cells using CoraLite® Plus 488 PPA1 antibody (CL488-68178, Clone: 1D11F4) at dilution of 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PPA1 Antibody in Immunocytochemistry (ICC/IF)
          PPA1 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-68178
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1D11F4
          Immunogen
          PPA1 fusion protein Ag7049
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MSGFSTEER AAPFSLEYRV FLKNEKGQYI SPFHDIPIYA DKDVFHMVVE VPRWSNAKME IATKDPLNPI KQDVKKGKLR YVANLFPYKG YIWNYGAIPQ TWEDPGHNDK HTGCCGDNDP IDVCEIGSKV CARGEIIGVK VLGILAMIDE GETDWKVIAI NVDDPDAANY NDINDVKRLK PGYLEATVDW FRRYKVPDGK PENEFAFNAE FKDKDFAIDI IKSTHDHWKA LVTKKTNGKG ISCMNTTLSE SPFKCDPDAA RAIVDALPPP CESACTVPTD VDKWFHHQKN
          Target Information
          The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.
          Bioinformatics
          Protein Aliases: cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; inorganic diphosphatase 1; Inorganic pyrophosphatase; IOPPP; iPPase; PPase; PPase-1; pyrophosphatase (inorganic) 1; pyrophosphatase 1; Pyrophosphate phospho-hydrolase; RP11-367H5.1
          View more View less
          Gene Aliases: HEL-S-66p; IOPPP; PP; PP1; PPA1; Pyp; SID6-8061
          View more View less
          UniProt ID: (Human) Q15181
          View more View less
          Entrez Gene ID: (Human) 5464, (Rat) 294504
          View more View less
          Function(s)
          magnesium ion binding inorganic diphosphatase activity pyrophosphatase activity hydrolase activity metal ion binding pyrophosphatase
          Process(es)
          phosphate-containing compound metabolic process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline