    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CAPZB Antibodies
          Proteintech
          CAPZB Monoclonal Antibody (1F9G2), CoraLite® Plus 488
          View all (15) CAPZB antibodies
          Tech Support
          Cite CAPZB Monoclonal Antibody (1F9G2), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CAPZB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CAPZB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CAPZB Antibody (CL488-68190) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed A431 cells using CoraLite® Plus 488 CAPZB antibody (CL488-68190, Clone: 1F9G2) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CAPZB Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-68190
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1F9G2
          Immunogen
          CAPZB fusion protein Ag19373
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: SDQQLDCAL DLMRRLPPQQ IEKNLSDLID LVPSLCEDLL SSVDQPLKIA RDKVVGKDYL LCDYNRDGDS YRSPWSNKYD PPLEDGAMPS ARLRKLEVEA NNAFDQYRDL YFEGGVSSVY LWDLDHGFAG VILIKKAGDG SKKIKGCWDS IHVVEVQEKS SGRTAHYKLT STVMLWLQTN KSGSGTMNLG GSLTRQMEKD ETVSDCSPHI ANIGRLVEDM ENKIRSTLNE IYFGKTKDIV NGL
          Target Information
          CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
          Bioinformatics
          Protein Aliases: Cap Z; capping actin protein of muscle Z-line beta subunit; capping protein (actin filament) muscle Z-line, beta; capping protein beta 1; CapZ beta; epididymis secretory sperm binding protein; F-actin capping protein beta subunit; F-actin-capping protein subunit beta; MGC104401; MGC129749; MGC129750; RP4-657E11.7; unnamed protein product
          View more View less
          Gene Aliases: 1700120C01Rik; AI325129; CAPB; CAPPB; Cappb1; CAPZ; CAPZB; CPB1; CPB2; CPbeat2; CPbeta1; CPbeta2
          View more View less
          UniProt ID: (Human) P47756, (Rat) Q5XI32, (Mouse) P47757
          View more View less
          Entrez Gene ID: (Human) 832, (Rat) 298584, (Mouse) 12345
          View more View less
          Function(s)
          actin binding protein binding cadherin binding molecular_function beta-tubulin binding actin filament binding actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein
          Process(es)
          cytoskeleton organization actin polymerization or depolymerization regulation of cell morphogenesis cell projection organization lamellipodium assembly actin cytoskeleton organization barbed-end actin filament capping actin filament capping cell morphogenesis biological_process regulation of lamellipodium assembly negative regulation of microtubule polymerization neuron projection development muscle fiber development negative regulation of filopodium assembly regulation of protein kinase C signaling
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline