Immunogen sequence: MAMSFEWPW QYRFPPFFTL QPNVDTRQKQ LAAWCSLVLS FCRLHKQSSM TVMEAQESPL FNNVKLQRKL PVESIQIVLE ELRKKGNLEW LDKSKSSFLI MWRRPEEWGK LIYQWVSRSG QNNSVFTLYE LTNGEDTEDE EFHGLDEATL LRALQALQQE HKAEIITVSD GRGVKFF
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
Protein Aliases: Dermal papilla-derived protein 9; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; HGNC:28122; hVps25; MGC10540; unnamed protein product; Vacuolar protein-sorting-associated protein 25
Gene Aliases: DERP9; EAP20; FAP20; VPS25
UniProt ID: (Human) Q9BRG1
Entrez Gene ID: (Rabbit) 103345024, (Human) 84313