          • Primary Antibodies ›
          • Mitofilin Antibodies
          Proteintech
          IMMT Monoclonal Antibody (2D2E8), CoraLite® Plus 488
          View all (20) Mitofilin antibodies
          Tech Support
          Cite IMMT Monoclonal Antibody (2D2E8), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          IMMT Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          IMMT Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          IMMT Antibody (CL488-68226) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using CoraLite® Plus 488 IMMT antibody (CL488-68226, Clone: 2D2E8) at dilution of 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          IMMT Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-68226
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2D2E8
          Immunogen
          IMMT fusion protein Ag0102
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: ACQLSGVTA AAQSCLCGKF VLRPLRPCRR YSTSGSSGLT TGKIAGAGLL FVGGGIGGTI LYAKWDSHFR ESVEKTIPYS DKLFEMVLGP AAYNVPLPKK SIQSGPLKIS SVSEVMKESK QPASQLQKQK GDTPASATAP TEAAQIISAA GDTLSVPAPA VQPEESLKTD HPEIGEGKPT PALSEEASSS SIRERPPEEV AARLAQQEKQ EQVKIESLAK SLEDALRQTA SVTLQAIAAQ NAAVQAVNAH SNILKAAMDN SEIAGEKKSA QWRTVEGALK ERRKAVDEAA DALLK
          Target Information
          Mitofilin is a transmembrane protein of the inner mitochondrial membrane and it is associated with a large multimeric protein complex of about 1200 kDa. Mitofilin has critical functions in mitochondrial morphology and mitochondrial fusion and fission, specifically in the formation of tubular cristae and cristae junctions. Abnormal mitochondrial morphology has been implicated in many human diseases such as myopathy, cardiomyopathy, rhabdomyosarcoma and Whartin's tumor.
          Bioinformatics
          Protein Aliases: Cell proliferation-inducing gene 4/52 protein; cell proliferation-inducing protein 52; heart muscle protein; IMMT; IMMT antibody; immt {ECO:0000312|EMBL:AAI05842.1, ECO:0000312|RGD:1310684}; inner mitochondrial transmembrane protein; MICOS complex subunit MIC60; mitochondrial contact site and cristae organizing system subunit 60; mitochondrial inner membrane organizing system 2; Mitochondrial inner membrane protein; mitochondrial inner membrane protein {ECO:0000250|UniProtKB:Q8CAQ8, ECO:0000312|EMBL:AAI05842.1}; Mitofilin; mitofilin {ECO:0000250|UniProtKB:Q8CAQ8}; motor protein; p87/89; p87/89 antibody; PIG4; PIG52; proliferation-inducing gene 4; unnamed protein product
          View more View less
          Gene Aliases: 1700082C19Rik; D830041H16Rik; HMP; IMMT; MIC60; MICOS60; MINOS2; P87; P87/89; P89; PIG4; PIG52
          View more View less
          UniProt ID: (Human) Q16891, (Rat) Q3KR86, (Mouse) Q8CAQ8
          View more View less
          Entrez Gene ID: (Human) 10989, (Rat) 312444, (Mouse) 76614
          View more View less
          Function(s)
          RNA binding protein binding molecular_function poly(A) RNA binding
          Process(es)
          response to cold mitochondrial calcium ion homeostasis inner mitochondrial membrane organization cristae formation neuron cellular homeostasis
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline