Immunogen sequence: MGARLSRRR LPADPSLALD ALPPELLVQV LSHVPPRSLV TRCRPVCRAW RDIVDGPTVW LLQLARDRSA EGRALYAVAQ RCLPSNEDKE EFPLCALARY CLRAPFGRNL IFNSCGEQGF RGWEVEHGGN GWAIEKNLTP VPGAPSQTCF VTSFEWCSKR QLVDLVMEGV WQELLDSAQI EICVADWWGA RENCGCVYQL RVRLLDVYEK EVVKFSASPD PVLQWTERGC RQVSHVFTNF GKGIRYVSFE QYGRDVSSWV GHYGALVTHS SVRVRIRLS
F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a third subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
Protein Aliases: F Box Gama 4; F-box only protein 17; F-box only protein 26; F-box protein FBG4; FBG4; FLJ11798; FLJ25205; FLJ58300; MGC9379; unnamed protein product
Gene Aliases: FBG4; FBX17; FBX26; FBXO17; FBXO26
UniProt ID: (Human) Q96EF6
Entrez Gene ID: (Human) 115290