    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • CARD11 Antibodies
          Proteintech
          CARD11 Monoclonal Antibody (3F5H6), CoraLite® Plus 488
          View all (18) CARD11 antibodies
          Tech Support
          Cite CARD11 Monoclonal Antibody (3F5H6), CoraLite® Plus 488
          • Antibody Testing Data (2)
          CARD11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CARD11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CARD11 Antibody (CL488-68269) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 CARD11 antibody (CL488-68269, Clone: 3F5H6) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CARD11 Antibody in Immunocytochemistry (ICC/IF)
          CARD11 Antibody in Flow Cytometry (Flow)
          CARD11 Monoclonal Antibody (3F5H6), CoraLite® Plus 488
          Product Details
          CL488-68269
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3F5H6
          Immunogen
          CARD11 fusion protein Ag16468
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: DTMYQDRHEW LCARVDPFTD HDLDMGTIPS YSRAQQLLLV KLQRLMHRGS REEVDGTHHT LRALRNTLQP EEALSTSDPR VSPRLSRASF LFGQLLQFVS RSENKYKRMN SNERVRIISG SPLGSLARSS LDATKLLTEK QEELDPESEL GKNLSLIPYS LVRAFYCERR RPVLFTPTVL AKTLVQRLLN SGGAMEFTIC KSDIVTRDEF LRRQKTETII YSREKNPNAF ECIAPANIEA VAAKNKHCLL EAGIGCTRDL IKSNIYPIVL FIRVCEKNIK RFRKLLPRPE TEEEFLRVCR LKEKELEALP CLYATVEPDM WGSVEELLRV VKDKIGEEQR KTIWVDEDQL
          Target Information
          The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein has a domain structure similar to that of CARD14 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10.
          Bioinformatics
          Protein Aliases: bcl10-interacting maguk protein 3; CARD-containing MAGUK protein 1; Carma 1; caspase recruitment domain family, member 11; Caspase recruitment domain-containing protein 11; MGC133069
          View more View less
          Gene Aliases: BENTA; BIMP3; CARD11; CARMA1; IMD11; PPBL
          View more View less
          UniProt ID: (Human) Q9BXL7
          View more View less
          Entrez Gene ID: (Human) 84433
          View more View less
          Function(s)
          guanylate kinase activity protein binding CARD domain binding
          Process(es)
          positive regulation of cytokine production stimulatory C-type lectin receptor signaling pathway positive regulation of B cell proliferation T cell costimulation Fc-epsilon receptor signaling pathway positive regulation of T cell proliferation regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling thymic T cell selection positive regulation of interleukin-2 biosynthetic process regulation of B cell differentiation regulation of T cell differentiation GMP metabolic process GDP metabolic process T cell receptor signaling pathway positive regulation of NF-kappaB transcription factor activity
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-55ff9ddd88-wrmqh:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0