    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CRTAC1 Antibodies
          Proteintech
          CRTAC1 Monoclonal Antibody (2A11E2), CoraLite® Plus 488
          View all (9) CRTAC1 antibodies
          Tech Support
          Cite CRTAC1 Monoclonal Antibody (2A11E2), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CRTAC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CRTAC1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CRTAC1 Antibody (CL488-68276) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using CoraLite® Plus 488 CRTAC1 antibody (CL488-68276, Clone: 2A11E2) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CRTAC1 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-68276
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rabbit, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2A11E2
          Immunogen
          CRTAC1 fusion protein Ag32813
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: VYGNWNGPH RLYLQMSTHG KVRFRDIASP KFSMPSPVRT VITADFDNDQ ELEIFFNNIA YRSSSANRLF RVIRREHGDP LIEELNPGDA LEPEGRGTGG VVTDFDGDGM LDLILSHGES MAQPLSVFRG NQGFNNNWLR VVPRTRFGAF ARGAKVVLYT KKSGAHLRII DGGSGYLCEM EPVAHFGLGK DEASSVEVTW PDGKMVSRNV ASGEMNSVLE ILYPRDEDTL QDPAPLECGQ GFSQQENGHC MDTNECIQFP FVCPRDKPVC VNTYGSYRCR TNKKCSRGYE PNEDGTACVG TLGQSPGPRP TTPTAAAATA AAAAAAGAAT AAPVLVDGDL NLGSVVKESC EPSC
          Target Information
          CRTAC1 encodes a glycosylated extracellular matrix protein that is found in the interterritorial matrix of articular deep zone cartilage. This protein is used as a marker to distinguish chondrocytes from osteoblasts and mesenchymal stem cells in culture. The presence of FG-GAP motifs and an RGD integrin-binding motif suggests that this protein may be involved in cell-cell or cell-matrix interactions. Copy number alterations in this gene have been observed in neurofibromatosis type 1-associated glomus tumors. Alternative splicing results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: 68 kDa chondrocyte-expressed protein; acidic secreted protein in cartilage; ASPIC; Cartilage acidic protein 1; CEP-68; chondrocyte expressed protein 68 kDa CEP-68; FLJ10320; Protein CRTAC1-B; unnamed protein product
          View more View less
          Gene Aliases: 2810454P21Rik; ASPIC; ASPIC1; AW047536; CEP-68; CEP68; CRTAC-1; CRTAC1; Crtac1B; LOTUS; W307
          View more View less
          UniProt ID: (Human) Q9NQ79, (Mouse) Q8R555
          View more View less
          Entrez Gene ID: (Rabbit) 100352703, (Human) 55118, (Pig) 100037974, (Mouse) 72832, (Rat) 171438
          View more View less
          Function(s)
          calcium ion binding protein binding
          Process(es)
          axonal fasciculation olfactory bulb development regulation of synapse assembly negative regulation of receptor binding
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline