FIGURE: 1 / 1
Immunogen sequence: MAELGLNEH HQNEVINYMR FARSKRGLRL KTVDSCFQDL KESRLVEDTF TIDEVSEVLN GLQAVVHSEV ESELINTAYT NVLLLRQLFA QAEKWYLKLQ TDISELENQE LLEQVAEFEK AEITSSNKKP ILDVTKPKLA PLNEGGTAEL LNEEILRLQE ENEKLKSRLK TIEIQATNAL DEKSKLEKAL QDLQLDQGNQ KDFIKAQDLS NLENTVAALK SEFQKTLNDK TENQKSLEEN LATAKHDLLR VREQLHMAEK ELEKKFQQTA AYRNMKEILT KKNDQIKDLR KRLAQYEPED
LZTFL1, also known as Leucine Zipper Transcription Factor-Like 1, is situated on the chromosomal region 3p21.3, which is frequently deleted in various cancers, including kidney cancer. It acts as a tumor suppressor by destabilizing AKT, a key protein in cell growth pathways, thus inhibiting tumor cell growth. Additionally, LZTFL1 has been implicated in the epithelial-mesenchymal transition in COVID-19, suggesting a role in viral response pathways. It is associated with Bardet-Biedl syndrome (BBS) and interacts with IFT27, a regulator of sperm flagellum formation, affecting spermatogenesis and male fertility. Knockout models show a phenotype of obesity, retinal degeneration, and abnormal cilia development, highlighting its role in normal ciliary function. The gene has two transcript isoforms and is part of a transcriptional map covering 250 kb, making it a subject of extensive genetic research.
Protein Aliases: Leucine zipper transcription factor-like protein 1; unnamed protein product
Gene Aliases: BBS17; LZTFL1
UniProt ID: (Human) Q9NQ48
Entrez Gene ID: (Human) 54585