    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • SERCA3 ATPase Antibodies
          Proteintech
          SERCA3 Monoclonal Antibody (1A2C2), CoraLite® Plus 488
          View all (6) SERCA3 ATPase antibodies
          Tech Support
          Cite SERCA3 Monoclonal Antibody (1A2C2), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SERCA3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SERCA3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SERCA3 Antibody (CL488-68287) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed HT-29 cells using CoraLite Plus 488 SERCA3 antibody (CL488-68287, Clone: 1A2C2 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SERCA3 Antibody in Immunocytochemistry (ICC/IF)
          SERCA3 Monoclonal Antibody (1A2C2), CoraLite® Plus 488
          Product Details
          CL488-68287
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1A2C2
          Immunogen
          SERCA3 fusion protein Ag4527
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: TRRMARKNAI VRSLPSVETL GCTSVICSDK TGTLTTNQMS VCRMFVVAEA DAGSCLLHEF TISGTTYTPE GEVRQGDQPV RCGQFDGLVE LATICALCND SALDYNEAKG VYEKVGEATE TALTCLVEKM NVFDTDLQAL SRVERAGACN TVIKQLMRKE FTLEFSRDRK SMSVYCTPTR PHPTGQGSKM FVKGAPESVI ERCSSVRVGS RTAPLTPTSR EQILAKIRDW GSGSDTLRCL ALATRDAPPR KEDMELDDCS KFVQYETDLT FVGCVGMLDP PRPEVAACIT RCYQAGIRVV MITGDNKGTA VAICRRLGIF GDTEDVAGKA YTGREFDDLS PEQQRQACRT ARCFARVEPA HKSRIVENLQ SFNEITAMTG DGVNDAPALK KAEIGIAMGS GTAVAKSAAE MVLSDDNFAS IVAAVEEGRA IYSNMKQFIR YLISSNVGEV VCIFLTAILG LPEALIPVQL LWVNLVTDGL PATALGFNPP DLDIMEKLPR SPREALISGW LFFRYLAIGV YVGLATVAAA TWWFVYDAEG PHINFYQLRN FLKCSEDNPL FAGIDCEVFE SRFPTTMALS VLVTIEMCNA LNSVSENQSL LRMPPWMNPW LLVAVAMSMA LHFLILLVPP LPLIFQVTPL SGRQWVVVLQ ISLPVILLDE ALKYLSRNHM HEEMSQK
          Target Information
          ATP dependent calcium pumps are responsible in part for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium ATPases (SERCA). The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific isoform expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. SERCA3 has been found to co-express with SERCA2b in platelets, mast cells, lymphoid cells, and epithelial cells. Due to this pattern of expression it is referred to as the non-muscle form of SERCA.
          Bioinformatics
          Protein Aliases: adenosine triphosphatase, calcium; ATPase, Ca(2+)-transporting, ubiquitous; ATPase, Ca++ transporting, ubiquitous; calcium ATPase; SERCA3f; Calcium pump 3; calcium-translocating P-type ATPase; sarco/endoplasmic reticulum Ca(2+)-ATPase 3-type Ca2+ pump; sarco/endoplasmic reticulum Ca2+ -ATPase; sarcoendoplasmic reticulum Ca2+ ATPase type 3; Sarcoplasmic/endoplasmic reticulum calcium ATPase 3; SERCA3; SERCA3D; SERCA3E; SR Ca(2+)-ATPase 3; unnamed protein product
          View more View less
          Gene Aliases: ATP2A3; SERCA3; SERCA3b
          View more View less
          UniProt ID: (Human) Q93084, (Rat) P18596, (Mouse) Q64518
          View more View less
          Entrez Gene ID: (Human) 489, (Rat) 25391, (Mouse) 53313
          View more View less
          Function(s)
          nucleotide binding calcium channel regulator activity calcium-transporting ATPase activity protein binding ATP binding cysteine-type endopeptidase activator activity involved in apoptotic process calcium ion transmembrane transporter activity ATPase activity calcium-dependent ATPase activity ion channel binding metal ion binding hydrolase activity primary active transporter
          Process(es)
          ion transport calcium ion transport cellular calcium ion homeostasis ion transmembrane transport intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress calcium ion transmembrane transport transport across blood-brain barrier calcium ion transport from cytosol to endoplasmic reticulum regulation of cardiac conduction metabolic process transport
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline