    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • MOCS2 Antibodies
          Proteintech
          MOCS2 Monoclonal Antibody (3H2D5), CoraLite® Plus 488
          View all (7) MOCS2 antibodies
          Tech Support
          Cite MOCS2 Monoclonal Antibody (3H2D5), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          MOCS2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MOCS2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          MOCS2 Antibody (CL488-68308) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using CoraLite® Plus 488 MOCS2 antibody (CL488-68308, Clone: 3H2D5) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MOCS2 Antibody in Immunocytochemistry (ICC/IF)
          MOCS2 Monoclonal Antibody (3H2D5), CoraLite® Plus 488
          Product Details
          CL488-68308
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3H2D5
          Immunogen
          MOCS2 fusion protein Ag6282
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MSSLEISSS CFSLETKLPL SPPLVEDSAF EPSRKDMDEV EEKSKDVINF TAEKLSVDEV SQLVISPLCG AISLFVGTTR NNFEGKKVIS LEYEAYLPMA ENEVRKICSD IRQKWPVKHI AVFHRLGLVP VSEASIIIAV SSAHRAASLE AVSYAIDTLK AKVPIWKKEI YEESSTWKGN KECFWASNS
          Target Information
          Eukaryotic molybdoenzymes use a unique molybdenum cofactor (MoCo) consisting of a pterin, termed molybdopterin, and the catalytically active metal molybdenum. MoCo is synthesized from precursor Z by the heterodimeric enzyme molybdopterin synthase. The large and small subunits of molybdopterin synthase are both encoded from this gene by overlapping open reading frames. The proteins were initially thought to be encoded from a bicistronic transcript. They are now thought to be encoded from monocistronic transcripts. Alternatively spliced transcripts have been found for this locus that encode the large and small subunits.
          Bioinformatics
          Protein Aliases: MOCO1-A; MOCO1-B; MOCS2; MOCS2A; MOCS2B; molybdenum cofactor; molybdenum cofactor biosynthesis protein E; molybdenum cofactor biosynthetic protein; Molybdenum cofactor synthesis protein 2 large subunit; Molybdenum cofactor synthesis protein 2 small subunit; Molybdenum cofactor synthesis protein 2A; Molybdenum cofactor synthesis protein 2B; Molybdopterin synthase catalytic subunit; Molybdopterin synthase sulfur carrier subunit; Molybdopterin-synthase large subunit; Molybdopterin-synthase small subunit; MPT synthase large subunit; Sulfur carrier protein MOCS2A; unnamed protein product
          View more View less
          Gene Aliases: MCBPE; MOCO1; MOCODB; MOCS2; MPTS
          View more View less
          UniProt ID: (Human) O96007
          View more View less
          Entrez Gene ID: (Human) 4338
          View more View less
          Function(s)
          nucleotide binding protein binding transferase activity molybdopterin synthase activity
          Process(es)
          Mo-molybdopterin cofactor biosynthetic process molybdopterin cofactor biosynthetic process
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline