Sequence of this protein is as follows: YEFPPYATNE IGKVTGLNRR ELGHGALAEK ALYPVIPRDF PFTIRVTSEV LESNGSSSMA SACGGSLALM DSGVPISSAV AGVAIGLVTK TDPEKGEIED YRLLTDILGI EDYNGDMDFK IAGTNKGITA LQADIKLPGI PIKIVMEAIQ QASVAKKEIL QIMNKTISKP RASRKENGPV VETVQVPLSK RAKFVGPGGY NLKKLQAETG VTISQVDEET FSVFAPTPSA MHEARDFITE ICKDDQEQQL EFGAVYTATI TEIRDTGVMV KLYPNMTAVL LHNTQLDQRK IKHPTALGLE VGQEIQVKYF GRDPADGRMR LSRKVLQSPA TTVVRTLNDR SSIVMGEPIS QSSSNSQ
PNPT1 (polyribonucleotide nucleotidyltransferase 1, mitochondirla) is a RNA-binding protein implicated in numerous RNA metabolic processes. It catalyzes the phosphorolysis of single-stranded polyribonucleotides processively in the 3'-to-5' direction. PNPT1 is a component of the mitochondrial degradosome (mtEXO) complex that degrades 3' overhang double-stranded RNA with a 3'-to-5' directionality in an ATP-dependent manner. It is required for correct processing and polyadenylation of mitochondrial mRNAs. It plays a role as a cytoplasmic RNA import factor, in mitochondrial morphogenesis and respiration, regulation of expression of the electron transport chain, regulation of the stability of specific mature miRNAs in melanoma cells, and RNA cell surveilance. Mutations of the gene can result in combined oxidative phosphorylation deficiency 13 (COXPD13) and deafness, autosomal recessive, 70 (DFNB70).
Protein Aliases: 3'-5' RNA exonuclease OLD35; DKFZp762K1914; old 35; Old-35; PNPase 1; PNPase old-35; Polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; Polyribonucleotide nucleotidyltransferase 1, mitochondrial
Gene Aliases: 1200003F12Rik; COXPD13; DFNB70; old-35; OLD35; PNPASE; PNPT1; Pnptl1; SCA25
UniProt ID: (Human) Q8TCS8, (Mouse) Q8K1R3
Entrez Gene ID: (Human) 87178, (Rat) 360992, (Mouse) 71701