    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SH3GLB2 Antibodies
          Proteintech
          SH3GLB2 Monoclonal Antibody (4B1A10), CoraLite® Plus 488
          View all (10) SH3GLB2 antibodies
          Tech Support
          Cite SH3GLB2 Monoclonal Antibody (4B1A10), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SH3GLB2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SH3GLB2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SH3GLB2 Antibody (CL488-68325) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using CoraLite® Plus 488 SH3GLB2 antibody (CL488-68325, Clone: 4B1A10) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SH3GLB2 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-68325
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rabbit, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4B1A10
          Immunogen
          SH3GLB2 fusion protein Ag8932
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MDFNMKKLA SDAGIFFTRA VQFTEEKFGQ AEKTELDAHF ENLLARADST KNWTEKILRQ TEVLLQPNPS ARVEEFLYEK LDRKVPSRVT NGELLAQYMA DAASELGPTT PYGKTLIKVA EAEKQLGAAE RDFIHTASIS FLTPLRNFLE GDWKTISKER RLLQNRRLDL DACKARLKKA KAAEAKATTV PDFQETRPRN YILSASASAL WNDEVDKAEQ ELRVAQTEFD RQAEVTRLLL EGISSTHVNH LRCLHEFVKS QTTYYAQCYR HMLDLQKQLG RFPGTFVGTT EPASPPLSST SPTTAAATMP VVPSVASLAP PGEASLCLEE VAPPASGTRK ARVLYDYEAA DSSELALLAD ELITVYSLPG MDPDWLIGER GNKKGKVPVT YLELLS
          Target Information
          SH3GLB2 (SH3 Domain Containing GRB2 Like, Endophilin B2) is a Protein Coding gene. Among its related pathways are Endocytosis. An important paralog of this gene is SH3GLB1. [GeneCards]
          Bioinformatics
          Protein Aliases: Endophilin-B2; retinoid receptor-induced gene 1; SH3 domain-containing GRB2-like protein B2; SH3-containing protein SH3GLB2; SH3-domain GRB2-like endophilin B2; sh3glb2 {ECO:0000312|EMBL:AAH87652.1}; stimulator of prostatic adenocarcinoma-specific T-cells-1
          View more View less
          Gene Aliases: KIAA1848; PP578; PP6569; PP9455; RRIG1; SH3GLB2; SPAS-1
          View more View less
          UniProt ID: (Human) Q9NR46, (Mouse) Q8R3V5
          View more View less
          Entrez Gene ID: (Human) 56904, (Rabbit) 100345396, (Pig) 100156344, (Mouse) 227700, (Rat) 311848
          View more View less
          Function(s)
          protein binding identical protein binding cadherin binding molecular_function
          Process(es)
          membrane organization biological_process
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline