Immunogen sequence: MDALVEDDI CILNHEKAHK RDTVTPVSIY SGDESVASHF ALVTAYEDIK KRLKDSEKEN SLLKKRIRFL EEKLIARFEE ETSSVGREQV NKAYHAYREV CIDRDNLKSK LDKMNKDNSE SLKVLNEQLQ SKEVELLQLR TEVETQQVMR NLNPPSSNWE VEKLSCDLKI HGLEQELELM RKECSDLKIE LQKAKQTDPY QEDNLKSRDL QKLSISSDNM QHAYWELKRE MSNLHLVTQV QAELLRKLKT STAIKKGKSL FTGNH
AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).
Protein Aliases: 5-azacytidine induced gene 2; NAK-associated protein 1; NF-kappa-B-activating kinase-associated protein 1; unnamed protein product
Gene Aliases: AZ2; NAP1; TILP
UniProt ID: (Human) Q9H6S1
Entrez Gene ID: (Human) 64343