Immunogen sequence: MSEVSCKKR DDYLEWPEYF MAVAFLSAQR SKDPNSQVGA CIVNSENKIV GIGYNGMPNG CSDDVLPWRR TAENKLDTKY PYVCHAELNA IMNKNLTDVK GCSMYVALFP CNECAKLIIQ AGIKEVIFMS DKYHDSDEAT AARLLFNMAG VTFRKFIPKC SKIVIDFDSI NSRPSQKLQ
The protein encoded by this gene catalyzes the deamination of dCMP to dUMP, the nucleotide substrate for thymidylate synthase. The encoded protein is allosterically activated by dCTP and inhibited by dTTP, and is found as a homohexamer. This protein uses zinc as a cofactor for its activity. Two transcript variants encoding different isoforms have been found for this gene.
Protein Aliases: dCMP deaminase; Deoxycytidylate deaminase; unnamed protein product
Gene Aliases: DCTD
UniProt ID: (Human) P32321
Entrez Gene ID: (Human) 1635