Sequence of this protein is as follows: QNVKHPVTGQ FLYQDSNWAS KVEKLTLDNA AFPFLAYSGI PAVSFCFCED TDYPYLGTTM DTYKELIERI PELNKVARAA AEVAGQFVIK LTHDVELNLD YERYNSQLLS FVRDLNQYRA DIKEMGLSLQ WLYSARGDFF RATSRLTTDF GNAEKTDRFV MKKLNDRVMR VEYHFLSPYV SPKESPFRHV FWGSGSHTLP ALLENLKLRK QNNGAFNETL FRNQLALATW TIQGAANALS G
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
Protein Aliases: CD antigen CD71; CD71; p90; T9; TR; transferrin receptor; Transferrin receptor 1; Transferrin receptor protein 1; unnamed protein product
Gene Aliases: CD71; IMD46; p90; T9; TFR; TFR1; TFRC; TR; TRFR
UniProt ID: (Human) Q1HE24
Entrez Gene ID: (Human) 7037