    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DEK Antibodies
          Proteintech
          DEK Recombinant Rabbit Monoclonal Antibody (230065H2), CoraLite® Plus 488
          View all (27) DEK antibodies
          Tech Support
          Cite DEK Recombinant Rabbit Monoclonal Antibody (230065H2), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          DEK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DEK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          DEK Antibody (CL488-82880) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite® Plus 488 DEK antibody (CL488-82880, Clone: 230065H2) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DEK Antibody in Immunocytochemistry (ICC/IF)
          DEK Antibody in Flow Cytometry (Flow)
          DEK Recombinant Rabbit Monoclonal Antibody (230065H2), CoraLite® Plus 488
          Product Details
          CL488-82880
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230065H2
          Immunogen
          DEK fusion protein Ag8225
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MSASAPAAEG EGTPTQPASE KEPEMPGPRE ESEEEEDEDD EEEEEEEKEK SLIVEGKREK KKVERLTMQV SSLQREPFTI AQGKGQKLCE IERIHFFLSK KKTDELRNLH KLLYNRPGTV SSLKKNVGQF SGFPFEKGSV QYKKKEEMLK KFRNAMLKSI CEVLDLERSG VNSELVKRIL NFLMHPKPSG KPLPKSKKTC SKGSKKERNS SGMARKAKRT KCPEILSDES SSDEDEKKNK EESSDDEDKE SEEEPPKKTA KREKPKQKAT SKSKKSVKSA NVKKADSSTT KKNQNSSKKE SESEDSSDDE PLIKKLKKPP TDEELKETIK KLLASANLEE VTMKQICKKV YENYPTYDLT ERKDFIKTTV KELIS
          Target Information
          DEK is a protein with one SAP domain. This protein binds to cruciform and superhelical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases.
          Bioinformatics
          Protein Aliases: DEK oncogene (DNA binding); Protein DEK; unnamed protein product
          View more View less
          Gene Aliases: D6S231E; DEK
          View more View less
          UniProt ID: (Human) P35659, (Rat) Q6AXS3
          View more View less
          Entrez Gene ID: (Human) 7913, (Rat) 306817
          View more View less
          Function(s)
          DNA binding RNA binding protein binding histone binding molecular_function poly(A) RNA binding chromatin/chromatin-binding, or -regulatory protein
          Process(es)
          chromatin organization chromatin remodeling regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter signal transduction viral genome replication positive regulation of transcription from RNA polymerase I promoter positive regulation of transcription from RNA polymerase II promoter positive regulation of transcription from RNA polymerase III promoter regulation of double-strand break repair regulation of double-strand break repair via nonhomologous end joining biological_process chromatin modification
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline