    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TBCD Antibodies
          Proteintech
          TBCD Recombinant Rabbit Monoclonal Antibody (230209F3), CoraLite® Plus 488
          View all (8) TBCD antibodies
          Tech Support
          Cite TBCD Recombinant Rabbit Monoclonal Antibody (230209F3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TBCD Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TBCD Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TBCD Antibody (CL488-82911-7) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed MCF-7 cells using CoraLite Plus 488 TBCD antibody (CL488-82911-7, Clone: 230209F3 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TBCD Antibody in Immunocytochemistry (ICC/IF)
          TBCD Recombinant Rabbit Monoclonal Antibody (230209F3), CoraLite® Plus 488
          Product Details
          CL488-82911-7
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230209F3
          Immunogen
          TBCD fusion protein Ag6310
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MALSDEPAAG GPEEEAEDET LAFGAALEAF GESAETRALL GRLREVHGGG AEREVALERF RVIMDKYQEQ PHLLDPHLEW MMNLLLDIVQ DQTSPASLVH LAFKFLYIIT KVRGYKTFLR LFPHEVADVE PVLDLVTIQN PKDHEAWETR YMLLLWLSVT CLIPFDFSRL DGNLLTQPGQ ARMSIMDRIL QIAESYLIVS DKARDAAAVL VSRFITRPDV KQSKMAEFLD WSLCNLARSS FQTMQGVITM DGTLQALAQI FKHGKREDCL
          Target Information
          Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
          Bioinformatics
          Protein Aliases: Beta-tubulin cofactor D; SSD-1; tfcD; Tubulin-folding cofactor D; Tubulin-specific chaperone D; unnamed protein product
          View more View less
          Gene Aliases: KIAA0988; PEBAT; PP1096; SSD-1; SSD1; TBCD; TFCD
          View more View less
          UniProt ID: (Human) Q9BTW9
          View more View less
          Entrez Gene ID: (Human) 6904
          View more View less
          Function(s)
          GTPase activator activity protein binding beta-tubulin binding chaperone binding
          Process(es)
          microtubule cytoskeleton organization mitotic cell cycle protein folding tubulin complex assembly post-chaperonin tubulin folding pathway negative regulation of cell-substrate adhesion negative regulation of microtubule polymerization adherens junction assembly cell morphogenesis involved in neuron differentiation bicellular tight junction assembly
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline