          • Primary Antibodies ›
          • NFX1 Antibodies
          Proteintech
          NFX1 Recombinant Rabbit Monoclonal Antibody (230211D9), CoraLite® Plus 488
          View all (8) NFX1 antibodies
          Tech Support
          Cite NFX1 Recombinant Rabbit Monoclonal Antibody (230211D9), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          NFX1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NFX1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          NFX1 Antibody (CL488-82924) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using CoraLite® Plus 488 NFX1 antibody (CL488-82924, Clone: 230211D9) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NFX1 Antibody in Immunocytochemistry (ICC/IF)
          NFX1 Antibody in Flow Cytometry (Flow)
          NFX1 Recombinant Rabbit Monoclonal Antibody (230211D9), CoraLite® Plus 488
          Product Details
          CL488-82924
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230211D9
          Immunogen
          NFX1 fusion protein Ag9448
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: TCARVHECDH PVYHSCHSEE KCPPCTFLTQ KWCMGKHEFR SNIPCHLVDI SCGLPCSATL PCGMHKCQRL CHKGECLVDE PCKQPCTTPR ADCGHPCMAP CHTSSPCPVT ACKAKVELQC ECGRRKEMVI CSEASSTYQR IAAISMASKI TDMQLGGSVE ISKLITKKEV HQARLECDEE CSALERKKRL AEAFHISEDS DPFNIRSSGS KFSDSLKEDA RKDLKFVSDV EKEMETLVEA VNKGKNSKKS HSFPPMNRDH RRIIHDLAQV YGLESVSYDS EPKRNVVVTA IRGKSVCPPT TLTGVLEREM QARPPPPIPH HRHQSDKNPG SSNLQKITKE PIIDYFDVQD
          Target Information
          MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. NFX1 is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma.MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. The protein encoded by this gene is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma. Three alternative splice variants, each of which encodes a different isoform, have been identified.
          Bioinformatics
          Protein Aliases: DKFZp779G2416; MGC20369; Nuclear transcription factor, X box-binding protein 1; Transcriptional repressor NF-X1; unnamed protein product; X-box binding 1
          View more View less
          Gene Aliases: NFX1; NFX2; TEG-42; Tex42
          View more View less
          UniProt ID: (Human) Q12986
          View more View less
          Entrez Gene ID: (Human) 4799
          View more View less
          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding nucleic acid binding DNA binding transcription factor activity, sequence-specific DNA binding RNA binding protein binding zinc ion binding transferase activity metal ion binding ubiquitin protein ligase activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter inflammatory response protein ubiquitination negative regulation of MHC class II biosynthetic process protein autoubiquitination
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline