    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PGM5 Antibodies
          Proteintech
          PGM5 Recombinant Rabbit Monoclonal Antibody (230206C7), CoraLite® Plus 488
          View all (13) PGM5 antibodies
          Tech Support
          Cite PGM5 Recombinant Rabbit Monoclonal Antibody (230206C7), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          PGM5 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          PGM5 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          PGM5 Antibody (CL488-82965) in Flow
          1x10^6 PC-3 cells were intracellularly stained with 0.8 µg Coralite® Plus 488 Pgm5 Recombinant Antibody (CL488-82965, Clone: 230206C7) (red), or 0.8 µg CoraLite® Plus 488-conjugated Rabbit IgG control Rabbit PolyAb (CL488-30000) (blue). Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PGM5 Antibody in Flow Cytometry (Flow)
          PGM5 Recombinant Rabbit Monoclonal Antibody (230206C7), CoraLite® Plus 488
          Product Details
          CL488-82965
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230206C7
          Immunogen
          PGM5 fusion protein Ag34399
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MEGSPIPVLT VPTAPYEDQR PAGGGGLRRP TGLFEGQRNY LPNFIQSVLS SIDLRDRQGC TMVVGSDGRY FSRTAIEIVV
          Target Information
          Component of adherens-type cell-cell and cell-matrix junctions. Lacks phosphoglucomutase activity.
          Bioinformatics
          Protein Aliases: Aciculin; PGM-RP; Phosphoglucomutase-like protein 5; Phosphoglucomutase-related protein; unnamed protein product
          View more View less
          Gene Aliases: PGM5; PGMRP
          View more View less
          UniProt ID: (Human) Q15124
          View more View less
          Entrez Gene ID: (Human) 5239
          View more View less
          Function(s)
          magnesium ion binding phosphoglucomutase activity structural molecule activity intramolecular transferase activity, phosphotransferases
          Process(es)
          carbohydrate metabolic process cell adhesion striated muscle tissue development myofibril assembly
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-l697b:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline