    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PCSK9 Antibodies
          Proteintech
          PCSK9 Recombinant Rabbit Monoclonal Antibody (230536E4), CoraLite® Plus 488
          View all (70) PCSK9 antibodies
          Tech Support
          Cite PCSK9 Recombinant Rabbit Monoclonal Antibody (230536E4), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          PCSK9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PCSK9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          PCSK9 Antibody (CL488-83004-4) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite Plus 488 PCSK9 antibody (CL488-83004-4, Clone: 230536E4 ) at dilution of 1:200, CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PCSK9 Antibody in Immunocytochemistry (ICC/IF)
          PCSK9 Recombinant Rabbit Monoclonal Antibody (230536E4), CoraLite® Plus 488
          Product Details
          CL488-83004-4
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230536E4
          Immunogen
          PCSK9 fusion protein Eg0640
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • CoraLite Plus 647
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: QDEDGDYEEL MLALPSQEDG LADEAAHVAT ATFRRCSKEA WRLPGTYIVV LMEETQRLQI EQTAHRLQTR AARRGYVIKV LHIFYDLFPG FLVKMSSDLL GLALKLPHVE YIEEDSFVFA QSIPWNLERI IPAWHQTEED RSPDGSSQVE VYLLDTSIQG AHREIEGRVT ITDFNSVPEE DGTRFHRQAS KCDSHGTHLA GVVSGRDAGV AKGTSLHSLR VLNCQGKGTV SGTLIGLEFI RKSQLIQPSG PLVVLLPLAG GYSRILNAAC RHLARTGVVL VAAAGNFRDD ACLYSPASAP EVITVGATNA QDQPVTLGTL GTNFGRCVDL FAPGKDIIGA SSDCSTCFMS QSGTSQAAAH VAGIVARMLS REPTLTLAEL RQRLIHFSTK DVINMAWFPE DQQVLTPNLV ATLPPSTHET GGQLLCRTVW SAHSGPTRTA TATARCAPEE ELLSCSSFSR SGRRRGDWIE AIGGQQVCKA LNAFGGEGVY AVARCCLVPR ANCSIHNTPA ARAGLETHVH CHQKDHVLTG CSFHWEVEDL SVRRQPALRS RRQPGQCVGH QAASVYASCC HAPGLECKIK EHGISGPSEQ VTVACEAGWT LTGCNVLPGA SLTLGAYSVD NLCVARVHDT ARADRTSGEA TVAAAICCRS RPSAKASWVQ
          Target Information
          PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
          Bioinformatics
          Protein Aliases: convertase subtilisin; hypercholesterolemia; NARC-1; neural apoptosis regulated convertase 1; Neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; Proprotein convertase 9; proprotein convertase PC9; Proprotein convertase subtilisin/kexin type 9; Subtilisin/kexin-like protease PC9
          View more View less
          Gene Aliases: AI415265; AI747682; FH3; HCHOLA3; Narc1; PC9; Pcsk9
          View more View less
          UniProt ID: (Mouse) Q80W65
          View more View less
          Entrez Gene ID: (Mouse) 100102
          View more View less
          Function(s)
          serine-type endopeptidase activity peptidase activity serine-type peptidase activity hydrolase activity sodium channel inhibitor activity low-density lipoprotein particle binding receptor inhibitor activity apolipoprotein binding very-low-density lipoprotein particle binding apolipoprotein receptor binding protein self-association poly(A) RNA binding low-density lipoprotein particle receptor binding very-low-density lipoprotein particle receptor binding serine protease
          Process(es)
          negative regulation of receptor recycling positive regulation of receptor internalization proteolysis lipid metabolic process triglyceride metabolic process phospholipid metabolic process apoptotic process lysosomal transport steroid metabolic process cholesterol metabolic process cellular response to starvation regulation of receptor activity negative regulation of low-density lipoprotein particle clearance protein processing protein autoprocessing neuron differentiation low-density lipoprotein receptor particle metabolic process low-density lipoprotein particle receptor catabolic process regulation of low-density lipoprotein particle receptor catabolic process positive regulation of low-density lipoprotein particle receptor catabolic process cellular response to insulin stimulus lipoprotein metabolic process cholesterol homeostasis regulation of neuron apoptotic process positive regulation of neuron apoptotic process negative regulation of receptor activity negative regulation of sodium ion transmembrane transporter activity
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline