    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SALL4 Antibodies
          Proteintech
          SALL4 Recombinant Rabbit Monoclonal Antibody (230284C4), CoraLite® Plus 488
          View all (35) SALL4 antibodies
          Tech Support
          Cite SALL4 Recombinant Rabbit Monoclonal Antibody (230284C4), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SALL4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SALL4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SALL4 Antibody (CL488-83039-4) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed Caco-2 cells using CoraLite Plus 488 SALL4 antibody (CL488-83039-4, Clone: 230284C4 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SALL4 Antibody in Immunocytochemistry (ICC/IF)
          SALL4 Recombinant Rabbit Monoclonal Antibody (230284C4), CoraLite® Plus 488
          Product Details
          CL488-83039-4
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230284C4
          Immunogen
          SALL4 fusion protein Ag16076
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MSRRKQAKPQ HINSEEDQGE QQPQQQTPEF ADAAPAAPAA GELGAPVNHP GNDEVASEDE ATVKRLRREE THVCEKCCAE FFSISEFLEH KKNCTKNPPV LIMNDSEGPV PSEDFSGAVL SHQPTSPGSK DCHRENGGSS EDMKEKPDAE SVVYLKTETA LPPTPQDISY LAKGKVANTN VTLQALRGTK VAVNQRSADA LPAPVPGANS IPWVLEQILC LQQQQLQQIQ LTEQIRIQVN MWASHALHSS GAGADTLKTL GSHMSQQVSA AVALLSQKAG SQGLSLDALK QAKLPHANIP SATSSLSPGL APFTLKPDGT RVLPNVMSRL PSALLPQAPG SVLFQSPFST
          Target Information
          Sall4 protein is encoded by the Sall4 gene located on chromosome 20 in humans and belongs to the Spalt-like protein family. It plays an important role in somatic stem cell maintenance and is studied as a stem cell biomarker. It is a zinc finger transcription factor that is upregulated during development and shown to interact with Oct4 and Sox2. It also forms a complex with Nanog to maintain the pluripotency of Embryonic Stem cells (ESCs). Several current research studies focus on its function as an oncogene and tumor biomarker. Some of the reported target genes of Sall4 include p53, Bmi-1, Bcl2 and XIAP.
          Bioinformatics
          Protein Aliases: dJ1112F19.1; Sal-like protein 4; Zinc finger protein 797; Zinc finger protein SALL4
          View more View less
          Gene Aliases: DRRS; HSAL4; IVIC; SALL4; ZNF797
          View more View less
          UniProt ID: (Human) Q9UJQ4
          View more View less
          Entrez Gene ID: (Human) 57167
          View more View less
          Function(s)
          RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding protein binding zinc ion binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development inner cell mass cell proliferation neural tube closure ventricular septum development regulation of transcription from RNA polymerase II promoter heart development tissue development neural tube development embryonic limb morphogenesis somatic stem cell population maintenance positive regulation of transcription from RNA polymerase II promoter
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline