    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • alpha COP Antibodies
          Proteintech
          COPA Recombinant Rabbit Monoclonal Antibody (230361G2), CoraLite® Plus 488
          View all (10) alpha COP antibodies
          Tech Support
          Cite COPA Recombinant Rabbit Monoclonal Antibody (230361G2), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          COPA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          COPA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          COPA Antibody (CL488-83089) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed A549 cells using CoraLite® Plus 488 COPA antibody (CL488-83089, Clone: 230361G2) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          COPA Antibody in Immunocytochemistry (ICC/IF)
          COPA Antibody in Flow Cytometry (Flow)
          COPA Recombinant Rabbit Monoclonal Antibody (230361G2), CoraLite® Plus 488
          Product Details
          CL488-83089
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230361G2
          Immunogen
          COPA fusion protein Ag34207
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: QLPVDHILAG SFETAMRLLH DQVGVIQFGP YKQLFLQTYA RGRTTYQALP CLPSMYGYPN RNWKDAGLKN GVPAVGLKLN DLIQRLQLCY QLTTVGKFEE AVEKFRSILL SVPLLVVDNK QEIAEAQQLI TICREYIVGL SVETERKKLP KETLEQQKRI CEMAAYFTHS NLQPVHMILV LRTALNLFFK LKNFKTAATF ARRLLELGPK PEVAQQTRKI LSACEKNPTD AYQLNYDMHN PFDICAASYR PIYRGKPVEK CPLSGACYSP EFKGQICRVT TVTEIGKDVI GLRISPLQFR
          Target Information
          In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by non-clathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms.
          Bioinformatics
          Protein Aliases: alpha coat protein; Alpha-coat protein; Alpha-COP; coatomer protein complex subunit alpha; Coatomer subunit alpha; HEP-COP; HEPCOP; proxenin; xenin
          View more View less
          Gene Aliases: AIAISD; AILJK; alpha-COP; AU040324; COPA; HEP-COP; xenin
          View more View less
          UniProt ID: (Human) P53621, (Mouse) Q8CIE6
          View more View less
          Entrez Gene ID: (Human) 1314, (Mouse) 12847
          View more View less
          Function(s)
          mRNA binding hormone activity structural molecule activity protein binding vesicle coat protein
          Process(es)
          intracellular protein transport ER to Golgi vesicle-mediated transport retrograde vesicle-mediated transport, Golgi to ER intra-Golgi vesicle-mediated transport signal transduction protein transport vesicle-mediated transport pancreatic juice secretion protein localization to axon protein localization to cell leading edge transport
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline