    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • NRF1 Antibodies
          Proteintech
          NRF1/nuclear respiratory factor 1 Recombinant Rabbit Monoclonal Antibody (230394B1), CoraLite® Plus 488
          View all (26) NRF1 antibodies
          Tech Support
          Cite NRF1/nuclear respiratory factor 1 Recombinant Rabbit Monoclonal Antibody (230394B1), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NRF1/nuclear respiratory factor 1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NRF1/nuclear respiratory factor 1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NRF1/nuclear respiratory factor 1 Antibody (CL488-83092) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite Plus 488 NRF1/nuclear respiratory factor 1 antibody (CL488-83092, Clone: 230394B1 ) at dilution of 1:200, CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NRF1/nuclear respiratory factor 1 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-83092
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230394B1
          Immunogen
          NRF1/nuclear respiratory factor 1 fusion protein Ag3172
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: LKYSTGRGKP GWGKESCKPI WWPEDIPWAN VRSDVRTEEQ KQRVSWTQAL RTIVKNCYKQ HGREDLLYAF EDQQTQTQAT ATHSIAHLVP SQTVVQTFSN PDGTVSLIQV GTGATVATLA DASELPTTVT VAQVNYSAVA DGEVEQNWAT LQGGEMTIQT TQASEATQAV ASLAEAAVAA SQEMQQGATV TMALNSEAAA HAVATLAEAT LQGGGQIVLS GETAAAVGAL TGVQDANGLF MADRAGRKWI LTDKATGLVQ IPVSMYQTVV TSLAQGNGPV QVAMAPVTTR ISDSAVTMDG QAVEVVTLEQ
          Target Information
          This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for "nuclear factor (erythroid-derived 2)-like 1" which has an official symbol of NFE2L1.
          Bioinformatics
          Protein Aliases: Alpha palindromic-binding protein; Alpha-pal; EWG; NRF-1; Nuclear respiratory factor 1; unnamed protein product
          View more View less
          Gene Aliases: ALPHA-PAL; C87038; D6Ertd415e; NRF1
          View more View less
          UniProt ID: (Human) Q16656, (Mouse) Q9WU00
          View more View less
          Entrez Gene ID: (Human) 4899, (Mouse) 18181
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding protein homodimerization activity transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding DNA-binding transcription factor
          Process(es)
          regulation of transcription from RNA polymerase II promoter positive regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter mitochondrion organization response to electrical stimulus
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0