Sequence of this protein is as follows: DVGEYNVFPR TLKWSKMNLT YRIVNYTPDM THSEVEKAFK KAFKVWSDVT PLNFTRLHDG IADIMISFGI KEHGDFYPFD GPSGLLAHAF PPGPNYGGDA HFDDDETWTS SSKGYNLFLV AAHEFGHSLG LDHSKDPGAL MFPIYTYTGK SHFMLPDDDV QGIQSLYGPG DEDPNPKHPK TPDKCDPSLS LDAITSLRGE TMIFKDRFFW RLHPQQVDAE LFLTKSFWPE LPNRIDAAYE HPSHDLIFIF RGRKFWALNG YDILEGYPKK ISELGLPKEV KKISAAVHFE DTGKTLLFSG NQVWRYDDTN HIMDKDYPRL IEEDFPGIGD KVDAVYEKNG YIYFFNGPIQ FEYSIWSNRI VRVMPANSIL WC
MMP13 (collagense 3) plays a role in the degradation of extracellular matrix proteins including fibrillar collagen, fibronectin, TNC, and ACAN. MMP13 cleaves tripal helical collagens, including type I, II, and III collagen - but has the highest activity with soluble type II collagen. It can degrade collagen type IV, XIV, and X. MMP13 plays a role in wound healing, tissue remodeling, cartilage degradation, bone development, bone mineralization, and ossification. It is required for normal embryonic bone development and ossificaiton. Mutations affecting the gene can lead to spondyloepimetaphyseal dysplasia Missouri type, metaphyseal anadysplasia 1, and metaphyseal dysplasia Sphar type.
Protein Aliases: Collagenase; Collagenase 3; Collagenase3; matrix metallo protease; matrix metalloproteinase 13 (collagenase 3); Matrix metalloproteinase-13; MMP; MMP-13; MMPs; Procollagenase 3; unnamed protein product
Gene Aliases: CLG3; MANDP1; MDST; MMP-13; MMP13
UniProt ID: (Human) P45452
Entrez Gene ID: (Human) 4322