    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TFAP4 Antibodies
          Proteintech
          TFAP4 Recombinant Rabbit Monoclonal Antibody (240031D5), CoraLite® Plus 488
          View all (17) TFAP4 antibodies
          Tech Support
          Cite TFAP4 Recombinant Rabbit Monoclonal Antibody (240031D5), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          TFAP4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TFAP4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          TFAP4 Antibody (CL488-83196-4) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed Hela cells using CoraLite® Plus 488 TFAP4 antibody (CL488-83196-4, Clone: 240031D5) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TFAP4 Antibody in Immunocytochemistry (ICC/IF)
          TFAP4 Antibody in Flow Cytometry (Flow)
          TFAP4 Recombinant Rabbit Monoclonal Antibody (240031D5), CoraLite® Plus 488
          Product Details
          CL488-83196-4
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240031D5
          Immunogen
          TFAP4 fusion protein Ag2633
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MEYFMVPTQK VPSLQHFRKT EKEVIGGLCS LANIPLTPET QRDQERRIRR EIANSNERRR MQSINAGFQS LKTLIPHTDG EKLSKAAILQ QTAEYIFSLE QEKTRLLQQN TQLKRFIQEL SGSSPKRRRA EDKDEGIGSP DIWEDEKAED LRREMIELRQ QLDKERSVRM MLEEQVRSLE AHMYPEKLKV IAQQVQLQQQ QEQVRLLHQE KLEREQQQLR TQLLPPPAPT HHPTVIVPAP PPPPSHHINV VTMGPSSVIN SVSTSRQNLD TIVQAIQHIE GTQEKQELEE EQRRAVIVKP VRSCPEAPTS DTASDSEASD SDAMDQSREE PSGDGELP
          Target Information
          Transcription factors of the basic helix-loop-helix-zipper(bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG.
          Bioinformatics
          Protein Aliases: Activating enhancer-binding protein 4; basic helix-loop-helix DNA-binding protein; bHLHc41; Class C basic helix-loop-helix protein 41; TFAP4; Transcription factor AP-4; transcription factor AP-4 (activating enhancer binding protein 4)
          View more View less
          Gene Aliases: AP-4; BHLHC41; TFAP4
          View more View less
          UniProt ID: (Human) Q01664
          View more View less
          Entrez Gene ID: (Human) 7023
          View more View less
          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding protein binding protein homodimerization activity histone deacetylase binding sequence-specific DNA binding protein dimerization activity E-box binding sequence-specific double-stranded DNA binding
          Process(es)
          regulation of transcription from RNA polymerase II promoter negative regulation of cell proliferation negative regulation of gene expression DNA damage response, signal transduction by p53 class mediator positive regulation of apoptotic process negative regulation of DNA binding negative regulation by host of viral transcription positive regulation by host of viral transcription negative regulation of cyclin-dependent protein serine/threonine kinase activity negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter protein-containing complex assembly cellular response to dexamethasone stimulus regulation of mitotic cell cycle phase transition
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline