FIGURE: 1 / 1
Immunogen sequence: RLPANTMQLF LDNNSIEGIP ENYFNVIPKV AFLRLNHNKL SDEGLPSRGF DVSSILDLQL SHNQLTKVPR ISAHLQHLHL DHNKIKSVNV SVICPSPSML PAERDSFSYG P
Keratocan (KTN), also known as keratan sulfate proteoglycan keratocan, is a protein that in humans is encoded by the KERA gene. It is mapped to 12q22. The protein encoded by this gene is a keratan sulfate proteoglycan that is involved in corneal transparency. Defects in this gene are a cause of autosomal recessive cornea plana 2 (CNA2). Keratan sulfate proteoglycans (KSPGs) are members of the small leucine-rich proteoglycan (SLRP) family. KSPGs, particularly keratocan, lumican and mimecan, are important to the transparency of the cornea.
Protein Aliases: Keratan sulfate proteoglycan keratocan; Keratocan; KTN; unnamed protein product
Gene Aliases: CNA2; KERA; KTN; SLRR2B
UniProt ID: (Human) O60938
Entrez Gene ID: (Human) 11081