    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • NUP85 Antibodies
          Proteintech
          NUP85 Recombinant Rabbit Monoclonal Antibody (240116A2), CoraLite® Plus 488
          View all (11) NUP85 antibodies
          Tech Support
          Cite NUP85 Recombinant Rabbit Monoclonal Antibody (240116A2), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NUP85 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NUP85 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NUP85 Antibody (CL488-83288) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite® Plus 488 NUP85 antibody (CL488-83288, Clone: 240116A2) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NUP85 Antibody in Immunocytochemistry (ICC/IF)
          NUP85 Recombinant Rabbit Monoclonal Antibody (240116A2), CoraLite® Plus 488
          Product Details
          CL488-83288
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240116A2
          Immunogen
          NUP85 fusion protein Ag7086
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: FLVTRLLYSN PTVKPIDLHY YAQSSLDLFL GGESSPEPLD NILLAAFEFD IHQVIKECSI ALSNWWFVAH LTDLLDHCKL LQSHNLYFGS NMREFLLLEY ASGLFAHPSL WQLGVDYFDY CPELGRVSLE LHIERIPLNT EQKALKVLRI CEQRQMTEQV RSICKILAMK AVRNNRLGSA LSWSIRAKDA AFATLVSDRF LRDYCERGCF SDLDLIDNLG PAMMLSDRLT FLGKYREFHR MYGEKRFADA ASLLLSLMTS RIAPRSFWMT LLTDALPLLE QKQVIFSAEQ TYELMRCLED LTSRRPVHGE SDTEQLQDDD IETTKVEMLR LSLARNLARA IIREGSLEGS
          Target Information
          Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP85 is part of one such subcomplex, Nup107-160.
          Bioinformatics
          Protein Aliases: 85 kDa nucleoporin; FLJ12549; FROUNT; Nuclear pore complex protein Nup85; nucleoporin 85kDa; Nucleoporin Nup75; Nucleoporin Nup85; PCN; PCNT; Pericentrin-1; unnamed protein product
          View more View less
          Gene Aliases: FROUNT; NPHS17; NUP75; NUP85; PCNT1
          View more View less
          UniProt ID: (Human) Q9BW27
          View more View less
          Entrez Gene ID: (Human) 79902
          View more View less
          Function(s)
          protein binding structural constituent of nuclear pore
          Process(es)
          mRNA export from nucleus protein import into nucleus nucleocytoplasmic transport chemotaxis protein transport lamellipodium assembly positive regulation of transcription, DNA-templated macrophage chemotaxis mRNA transport nephron development
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline