    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • STX1B Antibodies
          Proteintech
          STX1B Recombinant Rabbit Monoclonal Antibody (240156B9), CoraLite® Plus 488
          View all (4) STX1B antibodies
          Tech Support
          Cite STX1B Recombinant Rabbit Monoclonal Antibody (240156B9), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          STX1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          STX1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          STX1B Antibody (CL488-83298) in IHC (P)
          Immunofluorescent analysis of (4% PFA) fixed paraffin-embedded mouse brain tissue using CoraLite Plus 488 STX1B antibody (CL488-83298, Clone: 240156B9 ) at dilution of 1:200. Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          STX1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          STX1B Recombinant Rabbit Monoclonal Antibody (240156B9), CoraLite® Plus 488
          Product Details
          CL488-83298
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240156B9
          Immunogen
          STX1B fusion protein Ag10275
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MKDRTQELRS AKDSDDEEEV VHVDRDHFMD EFFEQVEEIR GCIEKLSEDV EQVKKQHSAI LAAPNPDEKT KQELEDLTAD IKKTANKVRS KLKAIEQSIE QEEGLNRSSA DLRIRKTQHS TLSRKFVEVM TEYNATQSKY RDRCKDRIQR QLEITGRTTT NEELEDMLES GKLAIFTDDI KMDSQMTKQA LNEIETRHNE IIKLETSIRE LHDMFVDMAM LVESQGEMID RIEYNVEHSV DYVERAVSDT KKAVKYQSKA RRK
          Target Information
          Potentially involved in docking of synaptic vesicles at presynaptic active zones. May mediate Ca(2+)-regulation of exocytosis acrosomal reaction in sperm.
          Bioinformatics
          Protein Aliases: STX1B-deltaTM; syntaxin 1B1; syntaxin 1B2; Syntaxin-1B; Syntaxin-1B1; Syntaxin-1B2
          View more View less
          Gene Aliases: GEFSP9; STX1B; STX1B1; STX1B2; Stx1bl; Syn1b
          View more View less
          UniProt ID: (Human) P61266, (Mouse) P61264
          View more View less
          Entrez Gene ID: (Human) 112755, (Mouse) 56216
          View more View less
          Function(s)
          SNARE binding receptor binding SNAP receptor activity protein binding protein kinase binding protein domain specific binding SNARE protein
          Process(es)
          positive regulation of neurotransmitter secretion neurotransmitter transport intracellular protein transport exocytosis vesicle docking involved in exocytosis regulation of gene expression regulation of synaptic vesicle priming negative regulation of neuron projection development synaptic vesicle exocytosis synaptic vesicle docking vesicle-mediated transport synaptic vesicle fusion to presynaptic active zone membrane vesicle docking calcium ion-regulated exocytosis of neurotransmitter regulation of synaptic activity spontaneous neurotransmitter secretion negative regulation of synaptic vesicle recycling positive regulation of spontaneous neurotransmitter secretion negative regulation of macropinocytosis positive regulation of excitatory postsynaptic potential transport protein transport regulation of exocytosis
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-794c4b9cd6-2x6nv:80/100.66.78.26:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline