    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • NCOA4 Antibodies
          Proteintech
          NCOA4 Recombinant Rabbit Monoclonal Antibody (240169H7), CoraLite® Plus 488
          View all (24) NCOA4 antibodies
          Tech Support
          Cite NCOA4 Recombinant Rabbit Monoclonal Antibody (240169H7), CoraLite® Plus 488
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          NCOA4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NCOA4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NCOA4 Antibody (CL488-83394-4) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite® Plus 488 NCOA4 antibody (CL488-83394-4, Clone: 240169H7) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NCOA4 Antibody in Immunocytochemistry (ICC/IF)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          NCOA4 Recombinant Rabbit Monoclonal Antibody (240169H7), CoraLite® Plus 488
          Product Details
          CL488-83394-4
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240169H7
          Immunogen
          NCOA4 fusion protein Ag15277
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MNTFQDQSGS SSNREPLLRC SDARRDLELA IGGVLRAEQQ IKDNLREVKA QIHSCISRHL ECLRSREVWL YEQVDLIYQL KEETLQQQAQ QLYSLLGQFN CLTHQLECTQ NKDLANQVSV CLERLGSLTL KPEDSTVLLF EADTITLRQT ITTFGSLKTI QIPEHLMAHA SSANIGPFLE KRGCISMPEQ KSASGIVAVP FSEWLLGSKP ASGYQAPYIP STDPQDWLTQ KQTLENSQTS SRACNFFNNV GGNLKGLENW LLKSEKSSYQ KCNSHSTTSS FSIEMEKVGD QELPDQDEMD LSDWLVTPQE SHKLRKPENG SRETSEKFKL LFQSYNVNDW LVKTDSCTNC QGNQPKGVEI ENLGNLKCLN DHLEAKKPLS TPSMVTEDWL VQNHQDPCKV
          Target Information
          Androgen receptor (AR) coactivator ARA70, also designated NCOA4, RFG and ELE1, is a putative co-activator that specifically enhances the activity of the androgen receptor. In human thyroid carcinomas, the Ret proto-oncogene fuses to ARA70 to form Ret/PTC3 by an intrachromosomal inversion of chromosome in vivo. ARA70 is expressed as two isoforms, ARA70a and ARA70b. The shorter variant, ARA70b, results from an internal 985-bp deletion. ARA70a is widely expressed, and its expression is highest in testis and adipose tissues, whereas ARA70b is solely expressed in the testis. ARA70a can function as a ligand-enhanced co-activator of PPARg in adipocytes. ARA70 may be involved in prostate carcinogenesis and ovarian cancer and may serve as a key mediator of estrogen-androgen synergism.
          Bioinformatics
          Protein Aliases: 70 kDa androgen receptor coactivator; 70 kDa AR-activator; androgen receptor-associated protein of 70 kDa; ELE1; ferritin cargo receptor NCOA4; NCoA-4; NCOA4; ret fused; RET-activating gene ELE1; unnamed protein product
          View more View less
          Gene Aliases: ARA70; ELE1; PTC3; RFG
          View more View less
          UniProt ID: (Human) Q13772
          View more View less
          Entrez Gene ID: (Human) 8031
          View more View less
          Function(s)
          transcription coactivator activity protein binding
          Process(es)
          protein targeting to lysosome cellular iron ion homeostasis autophagy male gonad development response to hormone erythrocyte differentiation intracellular estrogen receptor signaling pathway positive regulation of transcription, DNA-templated cellular response to estrogen stimulus cellular response to testosterone stimulus
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-hqqtp:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline