    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PTDSS1 Antibodies
          Proteintech
          PTDSS1 Recombinant Rabbit Monoclonal Antibody (240007G3), CoraLite® Plus 488
          View all (8) PTDSS1 antibodies
          Tech Support
          Cite PTDSS1 Recombinant Rabbit Monoclonal Antibody (240007G3), CoraLite® Plus 488
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          PTDSS1 Recombinant Rabbit Monoclonal Antibody (240007G3), CoraLite® Plus 488
          Product Details
          CL488-83421
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240007G3
          Immunogen
          PTDSS1 fusion protein Ag14170
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: TTFLCLYGMI WYAEHYGHRE KTYSECEDGT YSPEISWHHR KGTKGSEDSP PKHAGNNESH SSRRRNRHSK SKVTNGVGKK
          Target Information
          Catalyzes a base-exchange reaction in which the polar head group of phosphatidylethanolamine (PE) or phosphatidylcholine (PC) is replaced by L-serine. In membranes, PTDSS1 catalyzes mainly the conversion of phosphatidylcholine. Also converts, in vitro and to a lesser extent, phosphatidylethanolamine.
          Bioinformatics
          Protein Aliases: KIAA0024; PSS-1; ptdSer synthase 1; serine-exchange enzyme I; unnamed protein product
          View more View less
          Gene Aliases: LMHD; PSS1; PSSA
          View more View less
          UniProt ID: (Human) P48651
          View more View less
          Entrez Gene ID: (Human) 9791
          View more View less
          Function(s)
          transferase activity L-serine-phosphatidylethanolamine phosphatidyltransferase activity L-serine-phosphatidylcholine phosphatidyltransferase activity
          Process(es)
          phosphatidylserine biosynthetic process
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-ncdd4:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline