Sequence of this protein is as follows: KEAEKKVVIK SEAASPALPV ITKVETLSPE SALQGSPRSA ASTPAGSPDG SLPEHHAAAP NGLPGFSVEN IMTLRTSPPG GELSPGAGRA GLVVPPLALP YAAAPPAAYG QPCAQGLEAG AAGGYQCSMR AMSLYTGAER PAHMCVPPAL DEALSDHPSG PTSPLSALNL AAGQEGALAA TGHHHQHHGH HHPQAPPPPP APQPQPTPQP GAAAAQAASW YLNHSGDLNH LPGHTFAAQQ QTFPNVREMF NSHRLGIENS TLGESQVSGN ASCQLPYRST PPLYRHAAPY SYDCTKY*
Forkhead box protein C2 (FOXC2) is a transciptional activator and is characterized by a distinct DNA-binding forkhead domain. FOXC2 may be involved in the formation of special mesenchymal tissues. Mutations affecting the gene can cause lymphedema-distichiasis (LYD).
Protein Aliases: forkhead box C2 (MFH-1, mesenchyme forkhead 1); forkhead, Drosophila, homolog-like 14; forkhead-related protein FKHL14; mesenchyme fork head protein 1; mesenchyme forkhead 1; MFH-1,mesenchyme forkhead 1; transcription factor FKH-14
Gene Aliases: FKHL14; LD; MFH-1; MFH1
UniProt ID: (Human) Q99958
Entrez Gene ID: (Human) 2303