Immunogen sequence: HFYDRLQDAG SRWPELYLYS RADEVVLARD IERMVEARLA RRVLARSVDF VSSAHVSHLR DYPTYYTSLC VDFMRNCVRC
Protein Aliases: FLJ22353; novel DUF829 domain-containing protein; Nuclear envelope transmembrane protein 4; Transmembrane protein 53; unnamed protein product
Gene Aliases: CTDI; NET4; TMEM53
UniProt ID: (Human) Q6P2H8
Entrez Gene ID: (Human) 79639