Sequence of this protein is as follows: MLAAACQMFL GKTEAEIAHI ALETLEGHQR AIMAHMTVEE IYKDRQKFSE QVFKVASSDL VNMGISVVSY TLKDIHDDQD YLHSLGKART AQVQKDARIG EAEAKRDAGI REAKAKQEKV SAQYLSEIEM AKAQRDYELK KAAYDIEVNT RRAQADLAYQ LQVAKTKQQI EEQRVQVQVV ERAQQVAVQE QEIARREKEL EARVRKPAEA ERYKLERLAE AEKSQLIMQA EAEAASVRMR GEAEAFAIGA RARAEAEQMA KKAEAFQLYQ EAAQLDMLLE KLPQVAEEIS GPLTSANKIT LVSSGSGTMG AAKVTGEVLD ILTRLPESVE RLTGVSISQV NHKPLRTA
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
Protein Aliases: caveolae-associated protein; DADB-118P11.2; FLOT1; integral membrane component of caveolae; OTTHUMP00000164855; REG-2; reggie-2; unnamed protein product
Gene Aliases: Rareg; reggie-2
UniProt ID: (Human) O75955, (Mouse) O08917, (Rat) Q9Z1E1
Entrez Gene ID: (Human) 10211, (Mouse) 14251, (Rat) 64665