    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
          • Primary Antibodies ›
          • FAM127B Antibodies
          Proteintech
          FAM127B Recombinant Rabbit Monoclonal Antibody (240716B7), CoraLite® Plus 488
          View all (2) FAM127B antibodies
          Tech Support
          Cite FAM127B Recombinant Rabbit Monoclonal Antibody (240716B7), CoraLite® Plus 488
          • Antibody Testing Data (1)
          FAM127B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FAM127B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          FAM127B Antibody (CL488-83862-2) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite Plus 488 FAM127B antibody (CL488-83862-2, Clone: 240716B7 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FAM127B Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-83862-2
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240716B7
          Immunogen
          FAM127B fusion protein Ag13930
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MDGRVQLMKA LLAGPLRPAA RRWRNPIPFP ETFDGDTDRL PEFIVQTCSY MFVDENTFSN DALKVTFLIT RLTGPALQWV IPYIRKESPL LNDYRGFLAE MKRVFGWEED EDF
          Target Information
          The function of this protein remains unknown.
          Bioinformatics
          Protein Aliases: family with sequence similarity 127, member B; Mammalian retrotransposon derived protein 8A; Retrotransposon Gag-like protein 8A
          View more View less
          Gene Aliases: CXX1B; FAM127B; MAR8A; RTL8A
          View more View less
          UniProt ID: (Human) Q9BWD3
          View more View less
          Entrez Gene ID: (Human) 26071
          View more View less
          Function(s)
          protein binding
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline