    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • CD56 (NCAM) Antibodies
          Proteintech
          NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240936B10), CoraLite® Plus 488
          View all (186) CD56 (NCAM) antibodies
          Tech Support
          Cite NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240936B10), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NCAM1/CD56 Antibody (CL488-83883-5) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed Neuro-2a cells using CoraLite Plus 488 NCAM1/CD56 antibody (CL488-83883-5, Clone: 240936B10 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240936B10), CoraLite® Plus 488
          Product Details
          CL488-83883-5
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240936B10
          Immunogen
          Fusion Protein
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: LQVDIVPSQG EISVGESKFF LCQVAGDAKD KDISWFSPNG EKLSPNQQRI SVVWNDDDSS TLTIYNANID DAGIYKCVVT AEDGTQSEAT VNVKIFQKLM FKNAPTPQEF KEGEDAVIVC DVVSSLPPTI IWKHKGRDVI LKKDVRFIVL SNNYLQIRGI KKTDEGTYRC EGRILARGEI NFKDIQVIVN VPPTVQARQS IVNATANLGQ SVTLVCDADG FPEPTMSWTK DGEPIENEEE DDEKHIFSDD SSELTIRNVD KNDEAEYVCI AENKAGEQDA SIHLKVFAKP KITYVENQTA MELEEQVTLT CEASGDPIPS ITWRTSTRNI SSEEKTLDGH MVVRSHARVS SLTLKSIQYT DAGEYICTAS NTIGQDSQSM YLEFQYAPKL QGPVAVYTWE GNQVNITCEV FAYPSATISW FRDGQLLPSS NYSNIKIYNT PSASYLEVTP DSENDFGNYN CTAVNRIGQE SLEFILVQAD TPSSPSIDRV EPYSSTAQVQ FDEPEATGGV PILKYKAEWK SLGEESWHFK WYDAKEANME GIVTIMGLKP ETRYSVRLAA LNGKGLGEIS AATEFKTQPV REPSAPKLEG QMGEDGNSIK VNLIKQDDGG SPIRHYLVKY RALASEWKPE IRLPSGSDHV MLKSLDWNAE YEVYVVAENQ QGKSKAAHFV FRTSAQPTAI PANGSPTAGL ST
          Target Information
          CD56, also known as neural cell adhesion molecule (NCAM), is a highly glycosylated transmembrane glycoprotein of the immunoglobulin family. It plays a crucial role in cell adhesion, migration, axonal growth, pathfinding, and synaptic plasticity. CD56 is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa and is involved in homotypic adhesion of neural cells. It mediates interactions by binding extracellular matrix components such as laminin and integrins, with polysialic modification reducing CD56-mediated adhesion. In the hematopoietic system, CD56 is expressed on natural killer (NK) cells and a subset of T cells known as NKT cells. It is also found on most neuroectodermal-derived cell lines, tissues, and neoplasms, including retinoblastoma, medulloblastoma, astrocytomas, and neuroblastoma. CD56 serves as a widely used neuroendocrine marker with high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms, highlighting its importance in both neural and immune system functions.
          Bioinformatics
          Protein Aliases: CD-56; CD56; E NCAM; N CAM1; N-CAM-1; neural cell adhesion molecule; Neural cell adhesion molecule 1; sCD56; sNCAM; soluble CD56; soluble NCAM
          View more View less
          Gene Aliases: CD56; E-NCAM; Ncam; NCAM-1; Ncam1
          View more View less
          UniProt ID: (Mouse) P13595
          View more View less
          Entrez Gene ID: (Mouse) 17967
          View more View less
          Function(s)
          protein binding heparin binding phosphatase binding LRR domain binding identical protein binding
          Process(es)
          regulation of exocyst assembly cell adhesion cell surface receptor signaling pathway aging learning or memory response to lead ion response to fluoxetine thalamus development neuron projection development multicellular organismal response to stress homotypic cell-cell adhesion response to cocaine neuron development positive regulation of calcium-mediated signaling regulation of sensory perception of pain positive regulation of cardiac muscle cell proliferation negative regulation of cell death
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline