    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NEU2 Antibodies
          Proteintech
          NEU2 Recombinant Rabbit Monoclonal Antibody (240685C12), CoraLite® Plus 488
          View all (17) NEU2 antibodies
          Tech Support
          Cite NEU2 Recombinant Rabbit Monoclonal Antibody (240685C12), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NEU2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NEU2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NEU2 Antibody (CL488-83934-2) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed PC-12 cells using CoraLite Plus 488 NEU2 antibody (CL488-83934-2, Clone: 240685C12 ) at dilution of 1:200, CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NEU2 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-83934-2
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240685C12
          Immunogen
          NEU2 fusion protein Ag17386
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: VQWQAQEVVA QARLDGHRSM NPCPLYDAQT GTLFLFFIAI PGQVTEQQQL QTRANVTRLC QVTSTDHGRT WSSPRDLTDA AIGPAYREWS TFAVGPGHCL QLNDRARSLV VPAYAYRKLH PIQRPIPSAF CFLSHDHGRT WARGHFVAQD TLECQVAEVE TGEQRVVTLN ARSHLRARVQ AQSTNDGLDF QESQLVKKLV EPPPQGCQGS VISFPSPRSG PGSPAQWLLY THPTHSWQRA DLGAYLNPRP PAPEAWSEPV LLAKGSCAYS DLQSMGTGPD GSPLFGCLYE ANDYEEIVFL MFTLKQAFPA EYLPQ
          Target Information
          This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.
          Bioinformatics
          Protein Aliases: Cytosolic sialidase; N-acetyl-alpha-neuraminidase 2; sialidase 2 (cytosolic sialidase); Sialidase-2
          View more View less
          Gene Aliases: NEU2; SIAL2
          View more View less
          UniProt ID: (Human) Q9Y3R4
          View more View less
          Entrez Gene ID: (Human) 4759
          View more View less
          Function(s)
          exo-alpha-sialidase activity protein binding hydrolase activity hydrolase activity, acting on glycosyl bonds
          Process(es)
          glycoprotein catabolic process lipid metabolic process ganglioside catabolic process oligosaccharide catabolic process lipid catabolic process glycosphingolipid catabolic process
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline