Sequence of this protein is as follows: TQVTTQPKLG EVQQKCHQFC GWEGSGFPGA QPVSMDKQNI KLLDLKPYKV SWKADGTRYM MLIDGTNEVF MIDRDNSVFH VSNLEFPFRK DLRMHLSNTL LDGEMIIDRV NGQAVPRYLI YDIIKFNSQP VGDCDFNVRL QCIEREIISP RHEKMKTGLI DKTQEPFSVR NKPFFDICTS RKLLEGNFAK EVSHEMDGLI FQPTGKYKPG RCDDILKWKP PSLNSVDFRL KITRMGGEGL LPQNVGLLYV GGYERPFAQI KVTKELKQYD NKIIECKFEN NSWVFMRQRT DKSFPNAYNT AMAVCNSISN PVTKEMLFEF IDRCTAASQG QKRKHHLDPD TELMPPPPPK RPHPLT
Bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.
Protein Aliases: HCAP1; mRNA 5'-triphosphate monophosphatase; mRNA guanylyltransferase; RP3-488C13.1; unnamed protein product
Gene Aliases: CAP1A; hCAP; HCE; HCE1
UniProt ID: (Human) O60942
Entrez Gene ID: (Human) 8732