    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ETNPPL Isoform 1 Antibodies
          Proteintech
          AGXT2L1 Recombinant Rabbit Monoclonal Antibody (241119H5), CoraLite® Plus 488
          View all (8) ETNPPL Isoform 1 antibodies
          Tech Support
          Cite AGXT2L1 Recombinant Rabbit Monoclonal Antibody (241119H5), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          AGXT2L1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          AGXT2L1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          AGXT2L1 Antibody (CL488-83984-5) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using CoraLite Plus 488 AGXT2L1 antibody (CL488-83984-5, Clone: 241119H5 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          AGXT2L1 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-83984-5
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          241119H5
          Immunogen
          AGXT2L1 fusion protein Ag10584
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: IEISPYKFQK GKDVKKEFVH VAPTPDTYRG KYREDHADSA SAYADEVKKI IEDAHNSGRK IAAFIAESMQ SCGGQIIPPA GYFQKVAEYV HGAGGVFIAD EVQVGFGRVG KHFWSFQMYG EDFVPDIVTM GKPMGNGHPV ACVVTTKEIA EAFSSSGMEY FNTYGGNPVS CAVGLAVLDI IENEDLQGNA KRVGNYLTEL LKKQKAKHTL IGDIRGIGLF IGIDLVKDHL KRTPATAEAQ HIIYKMKEKR VLLSADGPHR NVLKIKPPMC FTEEDAKFMV DQLDRILTVL EEAMGTKTES VTSENTPCKT KMLKEAHIEL LRDSTTDSKE NPSRKRNGMC TDTHSLLSKR LKT
          Target Information
          Catalyzes the pyridoxal-phosphate-dependent breakdown of phosphoethanolamine, converting it to ammonia, inorganic phosphate and acetaldehyde.
          Bioinformatics
          Protein Aliases: Alanine--glyoxylate aminotransferase 2-like 1; alanine-glyoxylate aminotransferase 2-like 1; Ethanolamine-phosphate phospho-lyase; unnamed protein product
          View more View less
          Gene Aliases: 1300019H02Rik; AGXT2L1; AI195447; ETNPPL
          View more View less
          UniProt ID: (Human) Q8TBG4, (Mouse) Q8BWU8
          View more View less
          Entrez Gene ID: (Human) 64850, (Mouse) 71760
          View more View less
          Function(s)
          protein binding transaminase activity lyase activity pyridoxal phosphate binding ethanolamine-phosphate phospho-lyase activity catalytic activity transferase activity
          Process(es)
          phospholipid transfer to membrane response to food lipid homeostasis cellular response to glucocorticoid stimulus ceramide phosphoethanolamine catabolic process biological_process
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0