    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • UTP18 Antibodies
          Proteintech
          UTP18 Recombinant Rabbit Monoclonal Antibody (241240F1), CoraLite® Plus 488
          View all (11) UTP18 antibodies
          Tech Support
          Cite UTP18 Recombinant Rabbit Monoclonal Antibody (241240F1), CoraLite® Plus 488
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          UTP18 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UTP18 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          UTP18 Antibody (CL488-84035-3) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed A431 cells using CoraLite Plus 488 UTP18 antibody (CL488-84035-3, Clone: 241240F1 ) at dilution of 1:200, (CL594-phalloidin, red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          UTP18 Antibody in Immunocytochemistry (ICC/IF)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          UTP18 Recombinant Rabbit Monoclonal Antibody (241240F1), CoraLite® Plus 488
          Product Details
          CL488-84035-3
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          241240F1
          Immunogen
          UTP18 fusion protein Ag34662
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: RNRLRLEEDK PAVERCLEEL VFGDVENDED ALLRRLRGPR VQEHEDSGDS EVENEAKGNF PPQKKPVWVD EEDEDEEMVD MMNNRFRKDM MKNASESKLS KDNLKKRLKE EFQHAMGGVP AWAETTKRKT S
          Target Information
          UTP18 is involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.
          Bioinformatics
          Protein Aliases: similar to Homo sapiens CGI-48 protein mRNA with GenBank Accession Number AF151806; unnamed protein product; UTP18, small subunit (SSU) processome component, homolog; WD repeat domain 50; WD repeat-containing protein 50
          View more View less
          Gene Aliases: CGI-48; WDR50
          View more View less
          UniProt ID: (Human) Q9Y5J1
          View more View less
          Entrez Gene ID: (Human) 51096
          View more View less
          Function(s)
          RNA binding protein binding
          Process(es)
          rRNA processing maturation of SSU-rRNA ribosomal small subunit biogenesis
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline