    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TSEN34 Antibodies
          Proteintech
          TSEN34 Recombinant Rabbit Monoclonal Antibody (241155G4), CoraLite® Plus 488
          View all (10) TSEN34 antibodies
          Tech Support
          Cite TSEN34 Recombinant Rabbit Monoclonal Antibody (241155G4), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TSEN34 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TSEN34 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TSEN34 Antibody (CL488-84102-5) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed MCF-7 cells using CoraLite Plus 488 TSEN34 antibody (CL488-84102-5, Clone: 241155G4 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TSEN34 Antibody in Immunocytochemistry (ICC/IF)
          TSEN34 Recombinant Rabbit Monoclonal Antibody (241155G4), CoraLite® Plus 488
          Product Details
          CL488-84102-5
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          241155G4
          Immunogen
          TSEN34 fusion protein Ag35120
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: LMPEEARLLA EIGAVTLVSA PRPDSRHHSL ALTSFKRQQE ESFQEQSALA AEARETRRQE LLEKITEGQA AKKQKLEQAS GASSSQEAGS SQAAKEDETS DGQASGEQEE AGPS
          Target Information
          TSEN34 encodes a catalytic subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns from precursor tRNAs. The endonuclease complex is also associated with a pre-mRNA 3-prime end processing factor. A mutation in this gene results in the neurological disorder pontocerebellar hypoplasia type 2. Multiple alternatively spliced variants, encoding the same protein, have been identified.
          Bioinformatics
          Protein Aliases: CTD-3093M3.1; HsSen34; leukocyte receptor cluster (LRC) member 5; Leukocyte receptor cluster member 5; tRNA-intron endonuclease Sen34; tRNA-splicing endonuclease subunit Sen34; TSEN34 tRNA splicing endonuclease subunit; unnamed protein product
          View more View less
          Gene Aliases: LENG5; PCH2C; SEN34; SEN34L; TSEN34
          View more View less
          UniProt ID: (Human) Q9BSV6
          View more View less
          Entrez Gene ID: (Human) 79042
          View more View less
          Function(s)
          tRNA-intron endonuclease activity nucleic acid binding lyase activity
          Process(es)
          tRNA-type intron splice site recognition and cleavage tRNA splicing, via endonucleolytic cleavage and ligation mRNA processing tRNA processing
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline