    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Cytokeratin 14 Antibodies
          Proteintech
          Cytokeratin 14 Monoclonal Antibody (2G1E2), CoraLite® 594
          View all (88) Cytokeratin 14 antibodies
          Tech Support
          Cite Cytokeratin 14 Monoclonal Antibody (2G1E2), CoraLite® 594
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Cytokeratin 14 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cytokeratin 14 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Cytokeratin 14 Antibody (CL594-60320) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed A431 cells using CL594-60320 (Cytokeratin 14 antibody) at dilution of 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Cytokeratin 14 Antibody in Immunocytochemistry (ICC/IF)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Cytokeratin 14 Monoclonal Antibody (2G1E2), CoraLite® 594
          Product Details
          CL594-60320
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2G1E2
          Immunogen
          Recombinant protein
          3D Epitope / Immunogen
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite 488
          • CoraLite Plus 647
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Purification
          Protein G
          Storage buffer
          PBS with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LSSSQFSSGS QSSRDVTSSS RQIRTKVMDV HDGKVVSTHE QVLRTKN
          Target Information
          Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (ie. skin) and non-keratinizing cells (ie. mesothelial cells). At least 20 different CKs can be identified. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). Belonging to the type I subfamily of low molecular weight keratins and existing in combination with keratin 5, keratin 14 distinguishes stratified epithelial cells from simple epithelial cells and is useful in identification of squamous cell carcinomas. It is considered a prognostic marker in breast carcinomas. At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells.
          Bioinformatics
          Protein Aliases: cytokeratin 14; epidermal keratin type I; keratin 14, type I; keratin complex 1, acidic, gene 14; Koebner
          View more View less
          Gene Aliases: CK-14; CK14; EBS1; EBS1A; EBS1B; EBS1C; EBS1D; EBS3; EBS4; K14; Krt-1.14; Krt1-14; NFJ
          View more View less
          UniProt ID: (Human) P02533, (Mouse) Q61781
          View more View less
          Entrez Gene ID: (Human) 3861, (Mouse) 16664
          View more View less
          Function(s)
          structural molecule activity structural constituent of cytoskeleton protein binding structural constituent of epidermis keratin filament binding
          Process(es)
          morphogenesis of an epithelium epidermis development keratinocyte differentiation hair cycle intermediate filament organization intermediate filament bundle assembly positive regulation of keratinocyte differentiation response to radiation epithelial cell differentiation
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-l699q:80/100.66.79.160:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline