    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • Ninjurin 2 Antibodies
          Proteintech
          NINJ2 Monoclonal Antibody (5C5B2), CoraLite® 594
          View all (5) Ninjurin 2 antibodies
          Tech Support
          Cite NINJ2 Monoclonal Antibody (5C5B2), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NINJ2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          NINJ2 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NINJ2 Antibody (CL594-66202) in Flow
          1x10^6 U2OS cells were intracellularly stained with 0.4 µg Coralite594 Ninj2 Monoclonal Antibody (CL594-66202, Clone: 5C5B2) (red), or 0.4 µg CoraLite594 Mouse IgG2b isotype control (11B8C4) (CL594-66360-3, Clone: 11B8C4) (blue). Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF,00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NINJ2 Antibody in Flow Cytometry (Flow)
          NINJ2 Monoclonal Antibody (5C5B2), CoraLite® 594
          Product Details
          CL594-66202
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.40 µg per 10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5C5B2
          Immunogen
          NINJ2 fusion protein Ag5343
          3D Epitope / Immunogen
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MESARENIDL QPGSSDPRSQ PINLNHYATK KSVAESMLDV ALFMSNAMRL KAVLEQGPSS HYYTTLVTLI SLSLLLQVVI GVLLVVIARL NLNEVEKQWR LNQLNNAATI LVFFTVVINV FITAFGAHKT GFLAARASRN PL
          Target Information
          Ninjurin 2 (NINJ2) is a gene located on chromosome 12p13.2, encoding a member of the ninjurin family, which consists of cell surface proteins involved in promoting cell-cell adhesion and nerve regeneration. NINJ2 plays a pivotal role in neural repair processes, particularly in the peripheral nervous system, where it contributes to axonal growth and the formation of new neural connections following injury. This protein is expressed in various tissues, including the nervous system and endothelial cells, and is implicated in modulating inflammatory responses and cell-cell interactions. In addition to its role in the nervous system, NINJ2 is associated with vascular biology and immune cell migration, influencing processes such as angiogenesis and leukocyte transmigration. Altered expression or mutations in NINJ2 have been studied in the context of neurodegenerative diseases and stroke, where it may affect recovery and repair mechanisms. Research continues to explore NINJ2's potential as a therapeutic target for enhancing nerve regeneration and addressing vascular and inflammatory conditions.
          Bioinformatics
          Protein Aliases: FLJ56349; Nerve injury-induced protein 2; Ninjurin-2; unnamed protein product
          View more View less
          Gene Aliases: NINJ2
          View more View less
          UniProt ID: (Human) Q9NZG7
          View more View less
          Entrez Gene ID: (Human) 4815
          View more View less
          Function(s)
          protein binding cholesterol binding membrane destabilizing activity
          Process(es)
          cell adhesion neuron cell-cell adhesion nervous system development cytolysis tissue regeneration
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline