    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PD-1 (CD279) Antibodies
          Proteintech
          PD-1/CD279 Monoclonal Antibody (4H4D1), CoraLite® 594
          View all (332) PD-1 (CD279) antibodies
          Tech Support
          Cite PD-1/CD279 Monoclonal Antibody (4H4D1), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          PD-1/CD279 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          PD-1/CD279 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          PD-1/CD279 Antibody (CL594-66220) in IHC (PFA)
          Immunofluorescent analysis of (4% PFA) fixed paraffin-embedded human angioimmunoblastic T-cell lymphoma tissue using CoraLite®594 PD-1/CD279 antibody (CL594-66220, Clone: 4H4D1) at dilution of 1:200. Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PD-1/CD279 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          PD-1/CD279 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Product Details
          CL594-66220
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1:200
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4H4D1
          Immunogen
          PD-1/CD279 fusion protein Ag12470
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • CoraLite Plus 647
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MQIPQAPWP VVWAVLQLGW RPGWFLDSPD RPWNPPTFSP ALLVVTEGDN ATFTCSFSNT SESFVLNWYR MSPSNQTDKL AAFPEDRSQP GQDCRFRVTQ LPNGRDFHMS VVRARRNDSG TYLCGAISLA PKAQIKESLR AELRVTERRA EVPTAHPSPS PRPAGQFQTL VVGVVGGLLG SLVLLVWVLA VICSRAARGT IGARRTGQPL KEDPSAVPVF SVDYGELDFQ WREKTPEPPV PCVPEQTEYA TIVFPSGMGT SSPARRGSAD GPRSAQPLRP EDGHCSWPL
          Target Information
          Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "danger signals" such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
          Bioinformatics
          Protein Aliases: CD279; CD279; PD-1; hPD1; mPD-1; PD-1; PD1; alternatively spliced; PDCD1; programmed cell death 1 protein; Programmed cell death protein 1; programmed cell death protein 1-like; Protein PD-1; Protein PD1; sCD279; soluble CD279; systemic lupus erythematosus susceptibility 2; unnamed protein product
          View more View less
          Gene Aliases: AIMTBS; CD279; hPD-1; hPD-l; hSLE1; Ly101; PD-1; PD1; Pdc1; PDCD1; SLEB2
          View more View less
          UniProt ID: (Human) Q15116, (Mouse) Q02242
          View more View less
          Entrez Gene ID: (Human) 5133, (Rat) 301626, (Mouse) 18566
          View more View less
          Function(s)
          protein binding signaling receptor activity molecular_function
          Process(es)
          negative regulation of tolerance induction biological_process positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of T cell apoptotic process immune system process apoptotic process B cell apoptotic process adaptive immune response negative regulation of T cell mediated immune response to tumor cell negative regulation of B cell apoptotic process humoral immune response regulation of immune response negative regulation of immune response negative regulation of T cell activation negative regulation of multicellular organismal process regulatory T cell apoptotic process
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline